Assay Details
Target Gene Details
Entrez Gene ID: | 339230 |
Gene Name: | coiled-coil domain containing 137 |
Gene Aliases: |
RaRF |
Location: |
Chr.17:81666697-81673907 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCDC137 | NM_199287.2 | NP_954981.1 | ||
XM_011524738.2 | XP_011523040.1 | |||
XM_017024573.1 | XP_016880062.1 | |||
BC009369.2 | AAH09369.1 | |||
BM548065.1 | ||||
FJ649645.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv26223 | Chr.17:80659041 - 81736487 on Build GRCh38 | Gain+Loss | LOC101928855 NPLOC4 CEP131 MIR3186 HGS LOC105371921 RPTOR MIR6786 OXLD1 LOC100130370 TSPAN10 ACTG1 MIR4740 LOC100129503 CCDC137 SLC25A10 PDE6G BAIAP2-AS1 MRPL12 MIR3065 AATK-AS1 MIR657 LOC105371925 TEPSIN TMEM105 FSCN2 BAIAP2 LOC107985022 BAHCC1 LOC400627 C17orf89 LINC00482 CHMP6 AATK FAAP100 MIR338 MIR1250 LOC107985021 SLC38A10 ARL16 |
nsv428351 | Chr.17:81152631 - 83064073 on Build GRCh38 | Gain | FOXK2 NPLOC4 LOC101929511 MIR4525 MIR6787 MIR6786 SECTM1 PPP1R27 OXLD1 LOC100130370 ACTG1 SLC25A10 PDE6G PYCR1 RFNG FASN NARF AATK-AS1 ZNF750 CD7 PCYT2 TEPSIN BAHCC1 MCRIP1 RAC3 OGFOD3 C17orf89 GCGR MAFG-AS1 MAFG GPS1 C17orf62 LOC105376791 DUS1L HEXDC CEP131 MIR3186 HGS ARHGDIA LOC105371944 NOTUM TSPAN10 MIR4740 CCDC137 TBCD B3GNTL1 MRPL12 NPB CCDC57 SNORD134 ANAPC11 UTS2R ASPSCR1 FN3K LOC101929552 ALYREF RAB40B LOC105371925 STRA13 TMEM105 FSCN2 CSNK1D LRRC45 TEX19 LOC105376790 FN3KRP WDR45B DCXR SIRT7 P4HB LINC00482 AATK FAAP100 SLC16A3 MYADML2 SLC38A10 ARL16 |
nsv470620 | Chr.17:81516973 - 82064485 on Build GRCh38 | Loss | NPLOC4 HGS ARHGDIA MIR6786 NOTUM PPP1R27 OXLD1 TSPAN10 CCDC137 SLC25A10 PDE6G PYCR1 RFNG MRPL12 NPB ANAPC11 ASPSCR1 PCYT2 ALYREF STRA13 FSCN2 LRRC45 MCRIP1 RAC3 DCXR GCGR MAFG-AS1 SIRT7 P4HB MAFG GPS1 FAAP100 DUS1L MYADML2 ARL16 |
nsv952390 | Chr.17:81637175 - 81860124 on Build GRCh38 | Deletion | MCRIP1 HGS MIR6786 GCGR PPP1R27 OXLD1 TSPAN10 P4HB CCDC137 SLC25A10 PDE6G MRPL12 ARL16 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |