Assay Details
Target Gene Details
Entrez Gene ID: | 339230 |
Gene Name: | coiled-coil domain containing 137 |
Gene Aliases: |
RaRF |
Location: |
Chr.17:81666697-81673907 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCDC137 | NM_199287.2 | NP_954981.1 | ||
XM_011524738.2 | XP_011523040.1 | |||
BC009369.2 | AAH09369.1 | |||
BM548065.1 | ||||
FJ649645.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv26223 | Chr.17:80659041 - 81736487 on Build GRCh38 | Gain+Loss | MIR657 TEPSIN OXLD1 MIR3186 LOC400627 AATK MIR1250 MIR338 TMEM105 LINC00482 SLC25A10 SLC38A10 CHMP6 MIR4740 LOC105371921 FAAP100 MRPL12 LOC107985021 LOC101928855 ARL16 CCDC137 LOC107985022 C17orf89 LOC105371925 PDE6G CEP131 BAIAP2 ACTG1 BAHCC1 LOC100130370 BAIAP2-AS1 FSCN2 MIR6786 HGS RPTOR AATK-AS1 MIR3065 LOC100129503 TSPAN10 NPLOC4 |
nsv428351 | Chr.17:81152631 - 83064073 on Build GRCh38 | Gain | MIR3186 LOC105376790 AATK SECTM1 TEX19 TMEM105 ALYREF SLC25A10 SLC38A10 FN3K MIR4740 ARL16 CCDC57 MIR4525 LOC101929552 CCDC137 C17orf89 LOC105371925 PDE6G MAFG B3GNTL1 CEP131 GCGR ACTG1 BAHCC1 LOC100130370 FSCN2 FN3KRP WDR45B NARF NOTUM TBCD UTS2R HEXDC RAB40B FASN TSPAN10 CD7 RFNG OGFOD3 PCYT2 TEPSIN OXLD1 ANAPC11 RAC3 MCRIP1 DCXR PYCR1 LINC00482 LOC105376791 FAAP100 MRPL12 NPB LOC101929511 DUS1L ARHGDIA LRRC45 CSNK1D FOXK2 C17orf62 SIRT7 SNORD134 MIR6787 LOC105371944 STRA13 MIR6786 GPS1 ZNF750 HGS PPP1R27 ASPSCR1 SLC16A3 MYADML2 AATK-AS1 P4HB MAFG-AS1 NPLOC4 |
nsv470620 | Chr.17:81516973 - 82064485 on Build GRCh38 | Loss | PCYT2 OXLD1 ANAPC11 RAC3 MCRIP1 DCXR PYCR1 ALYREF SLC25A10 FAAP100 MRPL12 ARL16 NPB CCDC137 DUS1L PDE6G ARHGDIA LRRC45 MAFG GCGR SIRT7 FSCN2 STRA13 MIR6786 GPS1 HGS PPP1R27 ASPSCR1 NOTUM MYADML2 P4HB MAFG-AS1 TSPAN10 NPLOC4 RFNG |
nsv952390 | Chr.17:81637175 - 81860124 on Build GRCh38 | Deletion | GCGR OXLD1 MCRIP1 SLC25A10 MIR6786 HGS PPP1R27 MRPL12 ARL16 CCDC137 P4HB PDE6G TSPAN10 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |