Assay Details
Target Gene Details
Entrez Gene ID: | 314 |
Gene Name: | amine oxidase, copper containing 2 |
Gene Aliases: |
DAO2, RAO |
Location: |
Chr.17:42844592-42850707 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AOC2 | NM_001158.3 | NP_001149.2 | ||
NM_009590.2 | NP_033720.2 | |||
AF081363.1 | AAD39345.1 | |||
BC142641.1 | ||||
D88213.1 | BAA19001.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv483009 | Chr.17:42829470 - 43006887 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)