Assay Details
Target Gene Details
Entrez Gene ID: | 10750 |
Gene Name: | GRB2-related adaptor protein |
Gene Aliases: |
- |
Location: |
Chr.17:19020656-19051352 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GRAP | NM_006613.3 | NP_006604.1 | ||
XM_017024029.1 | XP_016879518.1 | |||
XM_017024030.1 | XP_016879519.1 | |||
BC035856.1 | AAH35856.1 | |||
BC063035.1 | AAH63035.1 | |||
BQ056077.1 | ||||
BT019981.1 | AAV38784.1 | |||
CR541941.1 | CAG46739.1 | |||
CR541966.1 | CAG46764.1 | |||
U52518.1 | AAC50541.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3892975 | Chr.17:19003716 - 19220206 on Build GRCh38 | Loss | SNORD3B-2 GRAPL SNORD3C GRAP LOC102724624 SNORD3D LOC388436 SLC5A10 SNORD3B-1 FAM83G LOC79999 SNORD3A |
nsv428336 | Chr.17:18927239 - 19288734 on Build GRCh38 | Gain+Loss | SNORD3B-2 GRAPL LOC102724624 SNORD3D LOC388436 EPN2 EPN2-IT1 SNORD3B-1 LOC79999 PRPSAP2 SNORD3A SNORD3C GRAP SLC5A10 FAM83G |
esv33572 | Chr.17:18919047 - 19291177 on Build GRCh38 | Gain | SNORD3B-2 GRAPL LOC102724624 SNORD3D LOC388436 EPN2 EPN2-IT1 SNORD3B-1 LOC79999 PRPSAP2 SNORD3A SNORD3C GRAP SLC5A10 FAM83G |
esv25927 | Chr.17:19024362 - 19238137 on Build GRCh38 | Gain+Loss | SNORD3B-2 GRAPL SNORD3C GRAP LOC102724624 SNORD3D LOC388436 EPN2 SNORD3B-1 LOC79999 SNORD3A |
dgv3116n100 | Chr.17:18957985 - 19190235 on Build GRCh38 | Gain | SNORD3B-2 GRAPL SNORD3C GRAP LOC102724624 SNORD3D SLC5A10 SNORD3B-1 FAM83G LOC79999 SNORD3A |
esv2758678 | Chr.17:18927303 - 19288734 on Build GRCh38 | Gain+Loss | SNORD3B-2 GRAPL LOC102724624 SNORD3D LOC388436 EPN2 EPN2-IT1 SNORD3B-1 LOC79999 PRPSAP2 SNORD3A SNORD3C GRAP SLC5A10 FAM83G |
nsv953842 | Chr.17:18947088 - 19049187 on Build GRCh38 | Deletion | GRAP SLC5A10 FAM83G |
nsv9504 | Chr.17:18389579 - 19238707 on Build GRCh38 | Gain+Loss | SNORD3B-2 GRAPL KRT17P5 LOC102724624 CCDC144B KRT16P1 ZNF286B GRAP EVPLL FLJ35934 LGALS9C FBXW10 KRT17P2 SNORD3D LOC388436 EPN2 SNORD3B-1 USP32P2 LOC79999 PRPSAP2 SNORD3A SNORD3C TBC1D28 TVP23B LOC101929141 SLC5A10 FAM106A TRIM16L FOXO3B FAM83G |
nsv960071 | Chr.17:19024288 - 19025116 on Build GRCh38 | Duplication | GRAP |
dgv895e212 | Chr.17:19019105 - 19248776 on Build GRCh38 | Loss | SNORD3B-2 GRAPL SNORD3C GRAP LOC102724624 SNORD3D LOC388436 EPN2 SLC5A10 SNORD3B-1 LOC79999 SNORD3A |
nsv833391 | Chr.17:18906887 - 19238887 on Build GRCh38 | Loss | SNORD3B-2 GRAPL LOC102724624 SNORD3D LOC388436 EPN2 SNORD3B-1 LOC79999 PRPSAP2 SNORD3A SNORD3C GRAP SLC5A10 FAM83G |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |