Assay Details
Target Gene Details
Entrez Gene ID: | 105371814 |
Gene Name: | uncharacterized LOC105371814 |
Gene Aliases: |
- |
Location: |
Chr.17:48874860-48908983 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC105371814 | NR_135674.1 | |||
CX873301.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833472 | Chr.17:48895840 - 49078381 on Build GRCh38 | Gain | IGF2BP1 SNF8 LOC105371814 ATP5G1 UBE2Z GIP |
nsv482969 | Chr.17:48843507 - 48994527 on Build GRCh38 | Loss | SNF8 LOC105371814 CALCOCO2 ATP5G1 UBE2Z GIP |