Assay Details
Target Gene Details
Entrez Gene ID: | 92736 |
Gene Name: | otopetrin 2 |
Gene Aliases: |
- |
Location: |
Chr.17:74922963-74933913 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
OTOP2 | NM_178160.2 | NP_835454.1 | ||
XM_011525479.1 | XP_011523781.1 | |||
AK000063.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833540 | Chr.17:74838971 - 75019365 on Build GRCh38 | Loss | TMEM104 FDXR FADS6 HID1-AS1 MRPL58 OTOP2 OTOP3 GRIN2C HID1 CDR2L USH1G |
nsv518292 | Chr.17:74810882 - 74982818 on Build GRCh38 | Loss | TMEM104 FDXR FADS6 HID1-AS1 OTOP2 OTOP3 GRIN2C HID1 USH1G |
nsv952367 | Chr.17:74751962 - 74961105 on Build GRCh38 | Deletion | SLC9A3R1 TMEM104 NAT9 FDXR FADS6 OTOP2 OTOP3 GRIN2C HID1 USH1G |
nsv576026 | Chr.17:74889500 - 74982818 on Build GRCh38 | Gain | FADS6 HID1-AS1 OTOP2 OTOP3 HID1 USH1G |
nsv1056788 | Chr.17:74922211 - 75028689 on Build GRCh38 | Gain | HID1-AS1 MRPL58 OTOP2 OTOP3 HID1 CDR2L USH1G |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |