Assay Details
Target Gene Details
Entrez Gene ID: | 51651 |
Gene Name: | peptidyl-tRNA hydrolase 2 |
Gene Aliases: |
BIT1, CFAP37, CGI-147, IMNEPD, PTH, PTH 2, PTH2 |
Location: |
Chr.17:59697306-59707598 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PTRH2 | NM_001015509.2 | NP_001015509.1 | ||
NM_016077.4 | NP_057161.1 | |||
XM_011524887.1 | XP_011523189.1 | |||
AF151905.1 | AAD34142.1 | |||
AK057033.1 | ||||
BC006807.1 | AAH06807.1 | |||
DA092491.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833501 | Chr.17:59627926 - 59779793 on Build GRCh38 | Loss | VMP1 PTRH2 CLTC |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |