Assay Details
Target Gene Details
Entrez Gene ID: | 10458 |
Gene Name: | BAI1 associated protein 2 |
Gene Aliases: |
BAP2, FLAF3, IRSP53 |
Location: |
Chr.17:81035147-81117432 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv26223 | Chr.17:80659041 - 81736487 on Build GRCh38 | Gain+Loss | LOC400627 ACTG1 LINC00482 NPLOC4 MIR1250 LOC107985021 LOC105371925 MIR338 LOC100130370 LOC107985022 CCDC137 CEP131 SLC25A10 MIR6786 MIR4740 BAIAP2-AS1 MIR657 C17orf89 TSPAN10 MRPL12 TEPSIN OXLD1 BAIAP2 AATK LOC100129503 AATK-AS1 RPTOR ARL16 PDE6G FAAP100 TMEM105 MIR3065 FSCN2 LOC101928855 MIR3186 HGS SLC38A10 BAHCC1 CHMP6 LOC105371921 |
nsv833564 | Chr.17:81014713 - 81164054 on Build GRCh38 | Loss | MIR3065 BAIAP2 AATK MIR1250 BAIAP2-AS1 MIR657 MIR338 |
nsv470619 | Chr.17:81032233 - 81349843 on Build GRCh38 | Loss | BAIAP2 AATK LINC00482 MIR1250 LOC105371925 AATK-AS1 MIR338 TMEM105 MIR3065 CEP131 BAIAP2-AS1 MIR657 SLC38A10 C17orf89 TEPSIN |
dgv3292n100 | Chr.17:81019582 - 81204920 on Build GRCh38 | Gain | MIR3065 CEP131 BAIAP2 AATK MIR1250 AATK-AS1 BAIAP2-AS1 MIR657 MIR338 |
nsv952388 | Chr.17:81034701 - 81137300 on Build GRCh38 | Deletion | MIR3065 BAIAP2 AATK MIR1250 BAIAP2-AS1 MIR657 MIR338 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |