Assay Details
Target Gene Details
Entrez Gene ID: | 339230 |
Gene Name: | coiled-coil domain containing 137 |
Gene Aliases: |
RaRF |
Location: |
Chr.17:81666697-81673907 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCDC137 | NM_199287.2 | NP_954981.1 | ||
XM_011524738.2 | XP_011523040.1 | |||
BC009369.2 | AAH09369.1 | |||
BM548065.1 | ||||
FJ649645.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv26223 | Chr.17:80659041 - 81736487 on Build GRCh38 | Gain+Loss | LOC107985021 MIR3065 HGS LOC100129503 MIR657 ACTG1 MIR1250 LOC101928855 CCDC137 TMEM105 FAAP100 MRPL12 SLC38A10 PDE6G NPLOC4 TEPSIN MIR338 LOC105371925 ARL16 BAHCC1 FSCN2 MIR3186 BAIAP2 OXLD1 LOC100130370 LOC105371921 SLC25A10 TSPAN10 MIR4740 LINC00482 AATK AATK-AS1 RPTOR BAIAP2-AS1 CEP131 C17orf89 LOC107985022 MIR6786 LOC400627 CHMP6 |
nsv428351 | Chr.17:81152631 - 83064073 on Build GRCh38 | Gain | NOTUM RAC3 GCGR ACTG1 TMEM105 OGFOD3 MRPL12 ARHGDIA MIR6787 LOC101929511 SECTM1 SLC38A10 PDE6G LOC105371925 ASPSCR1 LOC105371944 BAHCC1 FSCN2 SNORD134 MIR3186 ALYREF LOC105376791 PPP1R27 GPS1 MAFG-AS1 UTS2R FASN SLC25A10 LOC105376790 ZNF750 MCRIP1 FOXK2 STRA13 LRRC45 PYCR1 AATK-AS1 CSNK1D FN3K C17orf62 MAFG DUS1L HGS CCDC137 CCDC57 SIRT7 FAAP100 B3GNTL1 NARF HEXDC TBCD ANAPC11 MYADML2 NPLOC4 TEPSIN MIR4525 RAB40B ARL16 LOC101929552 P4HB OXLD1 LOC100130370 DCXR FN3KRP TSPAN10 PCYT2 MIR4740 LINC00482 AATK NPB TEX19 CEP131 C17orf89 MIR6786 RFNG CD7 WDR45B SLC16A3 |
nsv470620 | Chr.17:81516973 - 82064485 on Build GRCh38 | Loss | NOTUM DUS1L HGS RAC3 GCGR CCDC137 SIRT7 FAAP100 MRPL12 ARHGDIA ANAPC11 PDE6G MYADML2 NPLOC4 ASPSCR1 ARL16 FSCN2 ALYREF P4HB PPP1R27 OXLD1 GPS1 DCXR MAFG-AS1 SLC25A10 TSPAN10 MCRIP1 PCYT2 STRA13 LRRC45 NPB PYCR1 MIR6786 RFNG MAFG |
nsv952390 | Chr.17:81637175 - 81860124 on Build GRCh38 | Deletion | HGS P4HB PPP1R27 GCGR OXLD1 CCDC137 SLC25A10 TSPAN10 MRPL12 MCRIP1 PDE6G MIR6786 ARL16 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |