Assay Details
Target Gene Details
Entrez Gene ID: | 4025 |
Gene Name: | lactoperoxidase |
Gene Aliases: |
SPO |
Location: |
Chr.17:58238426-58268518 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LPO | NM_001160102.1 | NP_001153574.1 | ||
NM_006151.2 | NP_006142.1 | |||
NR_027647.1 | ||||
XM_011524808.2 | XP_011523110.1 | |||
AK131496.1 | ||||
AK300653.1 | ||||
AK303903.1 | ||||
AK308378.1 | ||||
AK310965.1 | ||||
BC107166.1 | AAI07167.1 | |||
BC107167.1 | AAI07168.1 | |||
BX487327.1 | ||||
BX648309.1 | ||||
DB230339.1 | ||||
EF579964.1 | ||||
U39573.1 | AAC50717.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv517229 | Chr.17:58176226 - 58373163 on Build GRCh38 | Gain+Loss | MIR142 TSPOAP1 TSPOAP1-AS1 RNF43 MIR4736 MKS1 LPO EPX SUPT4H1 MPO |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |