Assay Details
Target Gene Details
Entrez Gene ID: | 9368 |
Gene Name: | SLC9A3 regulator 1 |
Gene Aliases: |
EBP50, NHERF, NHERF-1, NHERF1, NPHLOP2 |
Location: |
Chr.17:74748612-74769360 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC9A3R1 | NM_004252.4 | NP_004243.1 | ||
AF015926.1 | AAC52084.1 | |||
AF036241.1 | AAC04572.1 | |||
AK026581.1 | ||||
AK094467.1 | ||||
AK128474.1 | ||||
AK312498.1 | ||||
BC001443.2 | AAH01443.1 | |||
BC003361.1 | AAH03361.1 | |||
BC011777.1 | AAH11777.1 | |||
BC049220.1 | AAH49220.1 | |||
BC053350.1 | AAH53350.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv952367 | Chr.17:74751962 - 74961105 on Build GRCh38 | Deletion | GRIN2C FADS6 FDXR OTOP2 SLC9A3R1 TMEM104 NAT9 OTOP3 USH1G HID1 |
nsv523611 | Chr.17:74755524 - 74767659 on Build GRCh38 | Loss | SLC9A3R1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |