Assay Details
Target Gene Details
Entrez Gene ID: | 226 |
Gene Name: | aldolase, fructose-bisphosphate A |
Gene Aliases: |
ALDA, GSD12, HEL-S-87p |
Location: |
Chr.16:30053090-30070420 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ALDOA | NM_000034.3 | NP_000025.1 | ||
XM_011545768.2 | XP_011544070.1 | |||
AK098778.1 | ||||
AK308495.1 | ||||
AK308748.1 | ||||
BC004333.1 | AAH04333.1 | |||
DB057572.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833187 | Chr.16:30051939 - 30173160 on Build GRCh38 | Loss | FAM57B YPEL3 TBX6 MAPK3 GDPD3 PPP4C ALDOA LOC101928595 |
nsv1160384 | Chr.16:30023148 - 30178660 on Build GRCh38 | Duplication | FAM57B YPEL3 DOC2A TBX6 MAPK3 GDPD3 PPP4C ALDOA LOC101928595 C16orf92 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |