Assay Details
Target Gene Details
Entrez Gene ID: | 8312 |
Gene Name: | axin 1 |
Gene Aliases: |
AXIN, PPP1R49 |
Location: |
Chr.16:287440-355226 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AXIN1 | NM_003502.3 | NP_003493.1 | ||
NM_181050.2 | NP_851393.1 | |||
NR_134879.1 | ||||
XM_011522682.2 | XP_011520984.1 | |||
XM_011522683.2 | XP_011520985.1 | |||
XM_011522684.2 | XP_011520986.1 | |||
XM_011522686.1 | XP_011520988.1 | |||
XM_017023743.1 | XP_016879232.1 | |||
XM_017023744.1 | XP_016879233.1 | |||
XM_017023745.1 | XP_016879234.1 | |||
XM_017023746.1 | XP_016879235.1 | |||
AB208876.1 | BAD92113.1 | |||
AF009674.1 | AAC51624.1 | |||
BC017447.1 | AAH17447.1 | |||
BC044648.1 | AAH44648.1 | |||
KF615847.1 | ||||
KF615848.1 | ||||
KF615849.1 | ||||
KF615850.1 | ||||
KF615851.1 | ||||
KF615852.1 | ||||
KF615853.1 | ||||
KF615854.1 | ||||
KU178413.1 | ||||
KU178414.1 | ||||
KU178415.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | MIR662 DECR2 MIR4717 RPL3L HBM UNKL TPSB2 LOC100134368 LMF1-AS1 SPSB3 RPS2 RAB26 NDUFB10 RPUSD1 MLST8 RHOT2 TSC2 LOC105371049 CCDC154 ERVK13-1 SNORD60 CAPN15 SNORA78 RNF151 HBA2 LOC106660606 LOC105371045 LINC00235 TMEM204 MSRB1 KCTD5 HN1L METTL26 JMJD8 ECI1 PTX4 NHLRC4 AMDHD2 PGP SNORA10 MIR3177 CCDC78 LUC7L IGFALS STUB1 NTHL1 MIR3677 MCRIP2 PKD1 ABCA3 LOC652276 PRSS29P MIR6859-4 ATP6V0C CEMP1 NPW FAM234A LOC100287175 RAB40C MIR6767 LOC105371046 RHBDL1 CACNA1H MIR6768 LINC00254 RAB11FIP3 FLJ42627 MIR3180-5 C16orf59 NPRL3 HBQ1 NARFL CRAMP1 RGS11 GFER LOC729652 WFIKKN1 EME2 SNHG19 NOXO1 LOC107984876 RNPS1 CCNF MSLN E4F1 SNHG9 PRR25 CLCN7 FBXL16 SNRNP25 AXIN1 DDX11L10 FAM173A MRPL28 TPSG1 SOX8 TPSD1 METRN PDIA2 MPG HS3ST6 CHTF18 UBE2I MIR3176 MIR1225 FAHD1 TMEM8A BAIAP3 GNG13 HBA1 TSR3 C1QTNF8 SSTR5 WASIR2 DNASE1L2 LOC105371047 WDR90 IFT140 RPL23AP5 NUBP2 MRPS34 NME3 HBZ SSTR5-AS1 BRICD5 POLR3K SYNGR3 MIR6511B1 PRR35 TRAF7 PIGQ C16orf91 TBC1D24 LOC105371038 LOC107987233 MEIOB HAGHL NTN3 GNPTG MIR5587 ABCA17P HAGH ARHGDIG LOC105371184 CASKIN1 MIR4516 RHBDF1 SNORA64 PDPK1 NME4 TBL3 ZNF598 MIR3178 TPSAB1 WDR24 MAPK8IP3 LOC101929280 MIR940 PRSS27 SLC9A3R2 LMF1 TELO2 |
esv3637564 | Chr.16:287941 - 370218 on Build GRCh38 | Gain | MRPL28 AXIN1 |
nsv952896 | Chr.16:273501 - 321500 on Build GRCh38 | Deletion | ARHGDIG RGS11 PDIA2 AXIN1 |
esv3637563 | Chr.16:263248 - 505590 on Build GRCh38 | Gain | RAB11FIP3 DECR2 MRPL28 FAM234A ARHGDIG RGS11 TMEM8A LOC100134368 RPL23AP5 NME4 PDIA2 AXIN1 |
nsv570951 | Chr.16:279671 - 322597 on Build GRCh38 | Loss | ARHGDIG PDIA2 AXIN1 |
nsv827477 | Chr.16:262247 - 297639 on Build GRCh38 | Gain | FAM234A ARHGDIG RGS11 PDIA2 AXIN1 |
nsv570947 | Chr.16:272935 - 297326 on Build GRCh38 | Loss | ARHGDIG RGS11 PDIA2 AXIN1 |
esv3637566 | Chr.16:296495 - 300305 on Build GRCh38 | Gain | AXIN1 |
dgv32n68 | Chr.16:245507 - 476226 on Build GRCh38 | Loss | RAB11FIP3 DECR2 MRPL28 FAM234A ARHGDIG RGS11 TMEM8A LOC100134368 RPL23AP5 NME4 PDIA2 AXIN1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |