Assay Details
Target Gene Details
Entrez Gene ID: | 100506083 |
Gene Name: | uncharacterized LOC100506083 |
Gene Aliases: |
- |
Location: |
Chr.16:70315640-70346747 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC100506083 | NR_039997.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833272 | Chr.16:70335866 - 70535820 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv2714670 | Chr.16:70331662 - 70370229 on Build GRCh38 | Deletion |
![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)