Assay Details
Target Gene Details
Entrez Gene ID: | 23199 |
Gene Name: | Gse1 coiled-coil protein |
Gene Aliases: |
CRHSP24, KIAA0182 |
Location: |
Chr.16:85169512-85676206 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GSE1 | XM_005255859.4 | XP_005255916.3 | ||
XM_005255860.3 | XP_005255917.3 | |||
XM_005255861.4 | XP_005255918.3 | |||
XM_017023084.1 | XP_016878573.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833312 | Chr.16:85158268 - 85327826 on Build GRCh38 | Loss | MIR5093 LINC00311 GSE1 |
dgv3059n100 | Chr.16:85068826 - 85219999 on Build GRCh38 | Gain | LOC400548 GSE1 FAM92B KIAA0513 |
nsv483044 | Chr.16:84166395 - 87066394 on Build GRCh38 | Gain | EMC8 MIR1910 COX4I1 ZDHHC7 LOC105376777 MIR6774 ADAD2 CRISPLD2 LOC102724344 IRF8 FLJ30679 ATP2C2 LOC101928557 LINC00917 FOXF1 FOXC2 LOC101928614 TLDC1 MIR5093 LOC400548 DNAAF1 GINS2 LOC440390 USP10 KCNG4 LOC146513 COTL1 GSE1 FENDRR LINC01082 KIAA0513 FOXL1 KLHL36 LOC101928474 TAF1C FOXC2-AS1 C16orf74 LINC00311 FAM92B LINC01081 MTHFSD LOC105371374 WFDC1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |