Assay Details
Target Gene Details
Entrez Gene ID: | 9091 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis class Q |
Gene Aliases: |
GPI1, c407A10.1 |
Location: |
Chr.16:569968-584136 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PIGQ | NM_004204.3 | NP_004195.2 | ||
NM_148920.2 | NP_683721.1 | |||
AB003723.1 | BAA24948.1 | |||
AF030177.1 | AAC32661.1 | |||
AK314361.1 | ||||
BC006377.2 | AAH06377.1 | |||
DA206420.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833113 | Chr.16:506513 - 628160 on Build GRCh38 | Loss | MIR3176 PRR35 PIGQ LOC107987233 NHLRC4 RAB11FIP3 LINC00235 MIR5587 LOC101929280 RAB40C CAPN15 |
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | SNORA64 LOC107987233 LOC106660606 LOC729652 MIR5587 RPL3L TBC1D24 DNASE1L2 PIGQ IFT140 TELO2 MIR6859-4 PRSS29P KCTD5 FAM173A FAM234A LINC00254 TMEM204 MIR662 RNF151 MIR6511B1 UBE2I ABCA17P RHBDF1 GNG13 TPSG1 NARFL RNPS1 HBQ1 SNRNP25 IGFALS CACNA1H LOC105371045 MIR3677 MCRIP2 ARHGDIG ATP6V0C MIR3180-5 RPL23AP5 METTL26 NME4 DECR2 LINC00235 CCDC154 MRPS34 RHOT2 AXIN1 EME2 LOC105371047 FLJ42627 WASIR2 MEIOB RAB26 TPSD1 PKD1 MIR6768 PRR25 CRAMP1 METRN POLR3K LMF1 RHBDL1 SPSB3 RGS11 PGP NOXO1 C16orf91 PDPK1 LOC105371046 ZNF598 RAB40C TBL3 LOC105371049 GNPTG SSTR5 RPS2 BRICD5 MIR3176 FBXL16 C16orf59 STUB1 WDR24 SNORD60 ECI1 ABCA3 TSC2 PRSS27 MSLN CLCN7 SNORA78 PRR35 NTHL1 HN1L MIR4717 TPSB2 LOC105371184 FAHD1 LOC101929280 NPRL3 NTN3 HBA1 CEMP1 LOC100287175 TPSAB1 PTX4 SNHG19 RAB11FIP3 LOC100134368 BAIAP3 HS3ST6 JMJD8 CCDC78 CASKIN1 CAPN15 E4F1 SSTR5-AS1 LOC105371038 MSRB1 HBA2 MIR6767 WFIKKN1 HBZ DDX11L10 RPUSD1 MAPK8IP3 NME3 MIR940 PDIA2 AMDHD2 HAGH SYNGR3 LOC107984876 HBM SNORA10 LOC652276 NDUFB10 MIR3177 CHTF18 NUBP2 MIR3178 GFER C1QTNF8 HAGHL CCNF TMEM8A MLST8 TRAF7 NPW WDR90 MIR1225 LMF1-AS1 SOX8 SLC9A3R2 MIR4516 TSR3 NHLRC4 LUC7L UNKL MPG ERVK13-1 MRPL28 SNHG9 |
esv2750209 | Chr.16:570921 - 571786 on Build GRCh38 | Deletion | PIGQ |
nsv471063 | Chr.16:520623 - 852322 on Build GRCh38 | Loss | MIR3176 FBXL16 STUB1 LOC107987233 WDR24 LINC00235 MIR5587 RHOT2 MSLN PRR35 PIGQ CHTF18 LOC105371184 LOC101929280 PRR25 METRN HAGHL FAM173A RHBDL1 LOC100287175 MIR662 RAB11FIP3 RAB40C GNG13 WDR90 NARFL JMJD8 CCDC78 CAPN15 LOC105371038 WFIKKN1 NHLRC4 MCRIP2 RPUSD1 METTL26 |
nsv526903 | Chr.16:451362 - 810673 on Build GRCh38 | Loss | MIR3176 FBXL16 STUB1 LOC107987233 WDR24 LINC00235 MIR5587 RHOT2 MSLN PRR35 PIGQ CHTF18 LOC105371184 LOC101929280 PRR25 METRN HAGHL FAM173A RHBDL1 LOC100287175 MIR662 RAB11FIP3 RAB40C GNG13 WDR90 NARFL JMJD8 CCDC78 CAPN15 LOC105371038 WFIKKN1 NHLRC4 MCRIP2 RPUSD1 METTL26 |
esv2750208 | Chr.16:570697 - 571406 on Build GRCh38 | Deletion | PIGQ |
nsv952898 | Chr.16:495001 - 1132900 on Build GRCh38 | Deletion | MIR3176 FBXL16 STUB1 LOC107987233 WDR24 LINC00235 MIR5587 LOC107984876 RHOT2 MSLN PRR35 PIGQ CHTF18 LOC105371184 LOC101929280 PRR25 C1QTNF8 METRN HAGHL FAM173A LMF1 RHBDL1 LOC100287175 MIR662 RAB11FIP3 RAB40C GNG13 WDR90 NARFL JMJD8 LMF1-AS1 CCDC78 SOX8 CAPN15 SSTR5-AS1 LOC105371038 WFIKKN1 NHLRC4 MCRIP2 RPUSD1 SSTR5 METTL26 |
dgv4823n54 | Chr.16:562698 - 662867 on Build GRCh38 | Loss | LOC105371038 PRR35 PIGQ LOC107987233 LOC100287175 WFIKKN1 NHLRC4 MCRIP2 LOC101929280 RAB40C WDR90 METTL26 |
nsv457304 | Chr.16:554554 - 611142 on Build GRCh38 | Gain | PRR35 PIGQ LOC107987233 NHLRC4 LOC101929280 RAB40C CAPN15 |
nsv1160325 | Chr.16:506925 - 836376 on Build GRCh38 | Duplication | MIR3176 FBXL16 STUB1 LOC107987233 WDR24 LINC00235 MIR5587 RHOT2 MSLN PRR35 PIGQ CHTF18 LOC105371184 LOC101929280 PRR25 METRN HAGHL FAM173A RHBDL1 LOC100287175 MIR662 RAB11FIP3 RAB40C GNG13 WDR90 NARFL JMJD8 CCDC78 CAPN15 LOC105371038 WFIKKN1 NHLRC4 MCRIP2 RPUSD1 METTL26 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |