Assay Details
Target Gene Details
Entrez Gene ID: | 23199 |
Gene Name: | Gse1 coiled-coil protein |
Gene Aliases: |
CRHSP24, KIAA0182 |
Location: |
Chr.16:85169512-85676206 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GSE1 | XM_005255859.4 | XP_005255916.3 | ||
XM_005255860.3 | XP_005255917.3 | |||
XM_005255861.4 | XP_005255918.3 | |||
XM_017023084.1 | XP_016878573.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833312 | Chr.16:85158268 - 85327826 on Build GRCh38 | Loss | MIR5093 LINC00311 GSE1 |
dgv3059n100 | Chr.16:85068826 - 85219999 on Build GRCh38 | Gain | LOC400548 KIAA0513 GSE1 FAM92B |
nsv483044 | Chr.16:84166395 - 87066394 on Build GRCh38 | Gain | DNAAF1 GINS2 LINC00917 LOC400548 FOXL1 TAF1C LOC102724344 USP10 LOC105371374 ADAD2 KIAA0513 FENDRR KLHL36 MIR5093 EMC8 FOXF1 C16orf74 FLJ30679 FOXC2 LINC01081 LOC440390 MIR1910 COTL1 LOC101928557 LOC105376777 LOC101928614 LINC00311 MIR6774 CRISPLD2 LOC101928474 COX4I1 MTHFSD KCNG4 LOC146513 WFDC1 TLDC1 FOXC2-AS1 GSE1 ATP2C2 IRF8 ZDHHC7 LINC01082 FAM92B |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |