Assay Details
Target Gene Details
Entrez Gene ID: | 9727 |
Gene Name: | RAB11 family interacting protein 3 |
Gene Aliases: |
CART1, Rab11-FIP3 |
Location: |
Chr.16:425621-522485 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RAB11FIP3 | NM_014700.3 | NP_055515.1 | ||
XM_005255713.3 | XP_005255770.1 | |||
XM_005255717.3 | XP_005255774.1 | |||
XM_005255718.3 | XP_005255775.1 | |||
XM_017023907.1 | XP_016879396.1 | |||
AB014565.1 | ||||
AF395731.1 | AAL12940.1 | |||
BC051360.1 | AAH51360.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | MIR1225 WFIKKN1 TMEM8A HBA2 SNHG9 EME2 TPSG1 C16orf59 IFT140 TELO2 CASKIN1 PKD1 C1QTNF8 AXIN1 HBZ TBL3 UBE2I RPL23AP5 CACNA1H CRAMP1 METTL26 HBM HS3ST6 SNORA10 CEMP1 RGS11 ATP6V0C MIR4516 TPSB2 LOC105371184 NARFL LOC105371047 TSC2 LOC105371038 CCNF FAM234A FLJ42627 NTHL1 RHBDL1 IGFALS LOC106660606 SSTR5-AS1 SPSB3 MRPL28 GNPTG TPSAB1 POLR3K PRSS27 NPRL3 RPS2 PGP RAB26 SNHG19 BRICD5 WASIR2 HAGHL HBA1 ZNF598 RHOT2 CLCN7 MIR662 PDPK1 LOC105371045 SNRNP25 FAHD1 MSLN TRAF7 BAIAP3 NTN3 NPW CHTF18 MIR5587 RNPS1 LOC105371049 LOC105371046 ERVK13-1 MIR3180-5 CAPN15 MRPS34 TSR3 LMF1 LUC7L RAB11FIP3 PTX4 MIR6767 HN1L FAM173A WDR90 PRSS29P SNORA78 DDX11L10 METRN NUBP2 MCRIP2 SYNGR3 TMEM204 LOC107987233 CCDC78 AMDHD2 DECR2 NHLRC4 HBQ1 GNG13 C16orf91 E4F1 SNORA64 FBXL16 NDUFB10 JMJD8 MEIOB UNKL RHBDF1 PRR25 ARHGDIG LOC729652 SNORD60 LOC101929280 PIGQ TBC1D24 LOC100134368 MIR3177 NME4 MIR3677 MLST8 GFER MIR6768 NME3 MIR3178 LINC00235 LOC652276 NOXO1 DNASE1L2 RPL3L ABCA17P RAB40C SSTR5 MIR940 STUB1 PRR35 MIR3176 LOC100287175 ABCA3 MPG MIR4717 LINC00254 MIR6859-4 KCTD5 RPUSD1 MIR6511B1 SOX8 SLC9A3R2 HAGH MSRB1 RNF151 LOC107984876 LMF1-AS1 TPSD1 PDIA2 ECI1 WDR24 MAPK8IP3 CCDC154 |
esv3637576 | Chr.16:439034 - 480097 on Build GRCh38 | Gain | RAB11FIP3 |
esv3637563 | Chr.16:263248 - 505590 on Build GRCh38 | Gain | AXIN1 ARHGDIG TMEM8A DECR2 RGS11 PDIA2 RPL23AP5 RAB11FIP3 LOC100134368 FAM234A MRPL28 NME4 |
nsv526903 | Chr.16:451362 - 810673 on Build GRCh38 | Loss | CAPN15 WFIKKN1 RAB11FIP3 LINC00235 FAM173A WDR90 RAB40C METRN STUB1 PRR35 MIR3176 MCRIP2 LOC100287175 HAGHL LOC107987233 METTL26 CCDC78 RHOT2 NHLRC4 RPUSD1 MIR662 GNG13 MSLN LOC105371184 FBXL16 NARFL JMJD8 PRR25 LOC105371038 CHTF18 LOC101929280 PIGQ MIR5587 WDR24 RHBDL1 |
nsv570954 | Chr.16:342461 - 530124 on Build GRCh38 | Loss | CAPN15 AXIN1 TMEM8A DECR2 RPL23AP5 RAB11FIP3 LOC100134368 LINC00235 MRPL28 NME4 |
dgv32n68 | Chr.16:245507 - 476226 on Build GRCh38 | Loss | AXIN1 ARHGDIG TMEM8A DECR2 RGS11 PDIA2 RPL23AP5 RAB11FIP3 LOC100134368 FAM234A MRPL28 NME4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |