Assay Details
Target Gene Details
Entrez Gene ID: | 9727 |
Gene Name: | RAB11 family interacting protein 3 |
Gene Aliases: |
CART1, Rab11-FIP3 |
Location: |
Chr.16:425621-522485 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RAB11FIP3 | NM_014700.3 | NP_055515.1 | ||
XM_005255713.3 | XP_005255770.1 | |||
XM_005255717.3 | XP_005255774.1 | |||
XM_005255718.3 | XP_005255775.1 | |||
XM_017023907.1 | XP_016879396.1 | |||
AB014565.1 | ||||
AF395731.1 | AAL12940.1 | |||
BC051360.1 | AAH51360.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | RPL23AP5 METTL26 FAHD1 CAPN15 ATP6V0C ARHGDIG TPSAB1 NME4 WFIKKN1 ERVK13-1 PKD1 HN1L LUC7L RPUSD1 CLCN7 LOC105371046 NUBP2 TPSG1 PDIA2 BAIAP3 PIGQ MIR940 NOXO1 TPSD1 LOC107987233 MIR3176 TSC2 NARFL HBM GFER UNKL TPSB2 PRR35 AMDHD2 LOC105371045 HBZ CCDC78 PDPK1 TELO2 LOC100287175 TBL3 E4F1 TMEM204 SLC9A3R2 LOC106660606 RGS11 NPW METRN UBE2I DNASE1L2 MEIOB IFT140 TRAF7 HAGHL TMEM8A C16orf91 SNHG9 TSR3 FAM234A MIR1225 MIR6768 HS3ST6 BRICD5 NTN3 NTHL1 C16orf59 JMJD8 MRPL28 EME2 GNPTG SYNGR3 TBC1D24 RNF151 LOC105371038 CEMP1 SNHG19 KCTD5 WDR24 CRAMP1 SOX8 C1QTNF8 NPRL3 RAB11FIP3 RPS2 CACNA1H RAB26 IGFALS MIR5587 MIR3178 RHBDF1 CCNF MRPS34 LOC100134368 MIR3177 RPL3L DDX11L10 ABCA3 RHOT2 MIR6511B1 LMF1 MSLN MIR4717 CCDC154 WDR90 LINC00254 LINC00235 RNPS1 LOC105371184 GNG13 LOC105371049 SNORA10 LOC105371047 SSTR5 LOC107984876 ABCA17P PRSS29P MLST8 DECR2 MIR6859-4 SNORD60 MIR3677 WASIR2 ZNF598 PRSS27 LMF1-AS1 MIR6767 LOC729652 PRR25 SNORA64 PGP NDUFB10 HBA2 SNRNP25 MPG ECI1 SPSB3 NME3 RHBDL1 MAPK8IP3 STUB1 HBQ1 SNORA78 FAM173A SSTR5-AS1 HBA1 POLR3K RAB40C CHTF18 HAGH MIR3180-5 CASKIN1 LOC101929280 MIR4516 MIR662 NHLRC4 MSRB1 AXIN1 PTX4 MCRIP2 LOC652276 FBXL16 FLJ42627 |
dgv4810n54 | Chr.16:380011 - 445662 on Build GRCh38 | Loss | RPL23AP5 LOC100134368 TMEM8A RAB11FIP3 NME4 DECR2 |
nsv9320 | Chr.16:427245 - 432556 on Build GRCh38 | Loss | RAB11FIP3 |
esv3637563 | Chr.16:263248 - 505590 on Build GRCh38 | Gain | RPL23AP5 LOC100134368 TMEM8A AXIN1 ARHGDIG RGS11 MRPL28 RAB11FIP3 NME4 FAM234A PDIA2 DECR2 |
esv3637574 | Chr.16:427856 - 430795 on Build GRCh38 | Loss | RAB11FIP3 |
nsv570956 | Chr.16:376432 - 437728 on Build GRCh38 | Gain | RPL23AP5 LOC100134368 TMEM8A RAB11FIP3 NME4 DECR2 |
nsv570954 | Chr.16:342461 - 530124 on Build GRCh38 | Loss | RPL23AP5 LOC100134368 LINC00235 TMEM8A AXIN1 CAPN15 MRPL28 RAB11FIP3 NME4 DECR2 |
nsv457303 | Chr.16:383623 - 445662 on Build GRCh38 | Loss | RPL23AP5 LOC100134368 RAB11FIP3 NME4 DECR2 |
dgv4809n54 | Chr.16:322597 - 451362 on Build GRCh38 | Loss | RPL23AP5 LOC100134368 TMEM8A AXIN1 MRPL28 RAB11FIP3 NME4 DECR2 |
esv2660167 | Chr.16:427853 - 430781 on Build GRCh38 | Deletion | RAB11FIP3 |
dgv32n68 | Chr.16:245507 - 476226 on Build GRCh38 | Loss | RPL23AP5 LOC100134368 TMEM8A AXIN1 ARHGDIG RGS11 MRPL28 RAB11FIP3 NME4 FAM234A PDIA2 DECR2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |