Assay Details
Target Gene Details
Entrez Gene ID: | 2653 |
Gene Name: | glycine cleavage system protein H |
Gene Aliases: |
GCE, NKH |
Location: |
Chr.16:81081947-81096375 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GCSH | NM_004483.4 | NP_004474.2 | ||
NR_033249.1 | ||||
XM_017023136.1 | XP_016878625.1 | |||
AK311978.1 | ||||
AV696698.1 | ||||
BF798008.1 | ||||
BG708388.1 | ||||
BT009827.1 | AAP88829.1 | |||
CN283805.1 | ||||
CN283809.1 | ||||
CR457072.1 | CAG33353.1 | |||
M69175.1 | AAA36011.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2751615 | Chr.16:81059494 - 81273994 on Build GRCh38 | Gain | PKD1L2 C16orf46 BCO1 GCSH |
nsv1062724 | Chr.16:81018707 - 81103402 on Build GRCh38 | Gain | CENPN PKD1L2 ATMIN C16orf46 GCSH |
nsv1067144 | Chr.16:81095693 - 81148036 on Build GRCh38 | Loss | PKD1L2 GCSH |
esv2422360 | Chr.16:81032428 - 81147122 on Build GRCh38 | Duplication | CENPN PKD1L2 ATMIN C16orf46 GCSH |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |