Assay Details
Target Gene Details
Entrez Gene ID: | 102467146 |
Gene Name: | MAF transcriptional regulator RNA |
Gene Aliases: |
linc-MAF-4, lincMAF4 |
Location: |
Chr.16:79721312-79770532 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MAFTRR | NR_104663.1 | |||
BX108785.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3639248 | Chr.16:79709018 - 79770282 on Build GRCh38 | Loss | MAFTRR |
esv3572208 | Chr.16:79668676 - 80101641 on Build GRCh38 | Gain | MAFTRR LINC01229 |
nsv833296 | Chr.16:79658339 - 79855830 on Build GRCh38 | Loss | MAFTRR LINC01229 |
nsv833297 | Chr.16:79698604 - 79886522 on Build GRCh38 | Gain | MAFTRR LINC01229 |
nsv573285 | Chr.16:79124285 - 80093429 on Build GRCh38 | Gain | MAFTRR LINC01229 WWOX LOC105371352 MAF |