Assay Details
Target Gene Details
Entrez Gene ID: | 9727 |
Gene Name: | RAB11 family interacting protein 3 |
Gene Aliases: |
CART1, Rab11-FIP3 |
Location: |
Chr.16:425621-522485 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RAB11FIP3 | NM_014700.3 | NP_055515.1 | ||
XM_005255713.3 | XP_005255770.1 | |||
XM_005255717.3 | XP_005255774.1 | |||
XM_005255718.3 | XP_005255775.1 | |||
XM_017023907.1 | XP_016879396.1 | |||
AB014565.1 | ||||
AF395731.1 | AAL12940.1 | |||
BC051360.1 | AAH51360.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | RAB26 RAB11FIP3 UBE2I GNG13 FLJ42627 PRR25 MIR662 ECI1 LINC00235 TELO2 WASIR2 SNHG9 LMF1 LOC106660606 ATP6V0C HBA2 LUC7L SNORD60 NPRL3 LINC00254 TSC2 RNF151 CCDC154 NPW ERVK13-1 PIGQ NDUFB10 HBZ LMF1-AS1 C16orf91 LOC105371038 MIR3677 DNASE1L2 MIR1225 MAPK8IP3 DECR2 TSR3 CCNF CASKIN1 TPSAB1 WDR90 LOC105371045 RPL3L MIR3180-5 MIR4516 MIR5587 FAM234A C1QTNF8 EME2 TBC1D24 MIR3177 ABCA17P RPUSD1 MSLN ABCA3 CHTF18 HBA1 LOC652276 MLST8 MIR6768 RAB40C CRAMP1 MSRB1 UNKL LOC105371047 MRPS34 E4F1 SNRNP25 LOC105371046 PRR35 HN1L TPSD1 PKD1 SNORA10 PRSS29P LOC105371184 LOC105371049 LOC101929280 SNHG19 FAHD1 MIR3178 TPSB2 MIR940 METRN AMDHD2 NARFL NTHL1 ZNF598 WDR24 TBL3 HBQ1 HBM MIR4717 HS3ST6 CLCN7 SPSB3 MEIOB POLR3K MIR6767 IGFALS MRPL28 RHOT2 BRICD5 NUBP2 STUB1 NHLRC4 SNORA64 TPSG1 RPL23AP5 RHBDL1 CCDC78 MIR3176 METTL26 PTX4 LOC729652 RNPS1 DDX11L10 LOC107984876 IFT140 TRAF7 LOC107987233 NOXO1 CACNA1H PGP LOC100134368 AXIN1 C16orf59 SLC9A3R2 GFER PDPK1 LOC100287175 FAM173A SNORA78 RPS2 MIR6859-4 RGS11 CAPN15 JMJD8 NME3 FBXL16 NME4 HAGH KCTD5 HAGHL TMEM8A SSTR5 MPG MCRIP2 WFIKKN1 TMEM204 SSTR5-AS1 GNPTG PDIA2 BAIAP3 ARHGDIG CEMP1 RHBDF1 SYNGR3 NTN3 PRSS27 MIR6511B1 SOX8 |
esv3637576 | Chr.16:439034 - 480097 on Build GRCh38 | Gain | RAB11FIP3 |
esv3637563 | Chr.16:263248 - 505590 on Build GRCh38 | Gain | RAB11FIP3 NME4 MRPL28 LOC100134368 TMEM8A RGS11 AXIN1 FAM234A RPL23AP5 DECR2 PDIA2 ARHGDIG |
nsv570954 | Chr.16:342461 - 530124 on Build GRCh38 | Loss | RAB11FIP3 NME4 MRPL28 LOC100134368 TMEM8A AXIN1 LINC00235 CAPN15 RPL23AP5 DECR2 |
dgv4809n54 | Chr.16:322597 - 451362 on Build GRCh38 | Loss | RAB11FIP3 NME4 MRPL28 LOC100134368 TMEM8A AXIN1 RPL23AP5 DECR2 |
dgv32n68 | Chr.16:245507 - 476226 on Build GRCh38 | Loss | RAB11FIP3 NME4 MRPL28 LOC100134368 TMEM8A RGS11 AXIN1 FAM234A RPL23AP5 DECR2 PDIA2 ARHGDIG |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |