Assay Details
Target Gene Details
Entrez Gene ID: | 22980 |
Gene Name: | transcription factor 25 |
Gene Aliases: |
FKSG26, Hulp1, NULP1, PRO2620, hKIAA1049 |
Location: |
Chr.16:89873583-89911384 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv573744 | Chr.16:89738447 - 89915950 on Build GRCh38 | Loss | LOC107984817 SPIRE2 FANCA LOC105371419 ZNF276 TCF25 |
esv2758671 | Chr.16:89804324 - 89901541 on Build GRCh38 | Loss | LOC107984817 SPIRE2 FANCA LOC105371419 TCF25 |
esv2762221 | Chr.16:89610380 - 90039318 on Build GRCh38 | Gain+Loss | AFG3L1P DPEP1 DEF8 LOC107984817 SPATA2L FANCA LOC105371419 DBNDD1 TUBB3 VPS9D1-AS1 CDK10 GAS8-AS1 VPS9D1 SPATA33 ZNF276 CHMP1A CENPBD1 SPIRE2 GAS8 MC1R TCF25 |
nsv573810 | Chr.16:89857498 - 90037251 on Build GRCh38 | Loss | AFG3L1P CENPBD1 DEF8 SPIRE2 GAS8 DBNDD1 TUBB3 MC1R GAS8-AS1 TCF25 |
nsv428331 | Chr.16:89216635 - 90185980 on Build GRCh38 | Gain+Loss | AFG3L1P DPEP1 DEF8 FANCA DBNDD1 VPS9D1-AS1 CDK10 VPS9D1 SPATA33 LOC105371414 CENPBD1 TUBB8P7 GAS8 URAHP CPNE7 MC1R LOC101927817 LOC107984817 FAM157C SPATA2L LOC105371419 TUBB3 ANKRD11 GAS8-AS1 RPL13 LOC100287036 ZNF276 CHMP1A SPIRE2 SPG7 SNORD68 ZNF778 PRDM7 TCF25 |
nsv573799 | Chr.16:89852186 - 90022379 on Build GRCh38 | Gain | AFG3L1P CENPBD1 DEF8 SPIRE2 GAS8 LOC105371419 DBNDD1 TUBB3 MC1R TCF25 |
nsv1066499 | Chr.16:89891957 - 90028891 on Build GRCh38 | Loss | AFG3L1P CENPBD1 DEF8 GAS8 DBNDD1 TUBB3 MC1R TCF25 |
nsv482951 | Chr.16:88633593 - 90228345 on Build GRCh38 | Loss | DPEP1 APRT DEF8 CTU2 VPS9D1-AS1 LOC105371414 CENPBD1 TUBB8P7 GAS8 LOC105371409 MIR4722 IL17C URAHP CPNE7 CBFA2T3 TRAPPC2L LOC101927817 LOC107984817 FAM157C PIEZO1 SPATA2L GAS8-AS1 RPL13 LOC101927793 SNAI3 SNORD68 MVD LINC00304 PRDM7 RNF166 ACSF3 AFG3L1P FANCA DBNDD1 CDK10 VPS9D1 GALNS SPATA33 LOC100129697 CDH15 LOC107987238 MC1R LOC400558 LOC339059 LOC105371419 TUBB3 ANKRD11 LOC100287036 ZNF276 LOC100289580 CHMP1A SLC22A31 SPIRE2 SPG7 ZNF778 SNAI3-AS1 PABPN1L CYBA CDT1 TCF25 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |