Assay Details
Target Gene Details
Entrez Gene ID: | 22980 |
Gene Name: | transcription factor 25 |
Gene Aliases: |
FKSG26, Hulp1, NULP1, PRO2620, hKIAA1049 |
Location: |
Chr.16:89873583-89911384 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv573744 | Chr.16:89738447 - 89915950 on Build GRCh38 | Loss | LOC107984817 FANCA LOC105371419 SPIRE2 ZNF276 TCF25 |
esv2758671 | Chr.16:89804324 - 89901541 on Build GRCh38 | Loss | LOC107984817 FANCA LOC105371419 SPIRE2 TCF25 |
esv2762221 | Chr.16:89610380 - 90039318 on Build GRCh38 | Gain+Loss | LOC107984817 CHMP1A VPS9D1-AS1 LOC105371419 SPIRE2 ZNF276 CENPBD1 TCF25 GAS8-AS1 DBNDD1 CDK10 GAS8 FANCA AFG3L1P TUBB3 DPEP1 SPATA33 DEF8 MC1R SPATA2L VPS9D1 |
nsv573810 | Chr.16:89857498 - 90037251 on Build GRCh38 | Loss | GAS8-AS1 DBNDD1 GAS8 AFG3L1P SPIRE2 TUBB3 CENPBD1 DEF8 MC1R TCF25 |
nsv428331 | Chr.16:89216635 - 90185980 on Build GRCh38 | Gain+Loss | LOC100287036 CHMP1A ZNF276 SNORD68 FAM157C SPG7 CPNE7 LOC101927817 ANKRD11 GAS8-AS1 GAS8 FANCA TUBB8P7 ZNF778 DPEP1 LOC105371414 DEF8 LOC107984817 RPL13 VPS9D1-AS1 LOC105371419 SPIRE2 CENPBD1 PRDM7 TCF25 URAHP DBNDD1 CDK10 AFG3L1P TUBB3 SPATA33 MC1R SPATA2L VPS9D1 |
nsv573799 | Chr.16:89852186 - 90022379 on Build GRCh38 | Gain | DBNDD1 GAS8 AFG3L1P LOC105371419 SPIRE2 TUBB3 CENPBD1 DEF8 MC1R TCF25 |
nsv9479 | Chr.16:89878586 - 89880246 on Build GRCh38 | Gain | TCF25 |
nsv482951 | Chr.16:88633593 - 90228345 on Build GRCh38 | Loss | LOC100287036 CHMP1A CTU2 LINC00304 SNAI3-AS1 LOC107987238 LOC400558 ANKRD11 RNF166 GAS8-AS1 LOC339059 TUBB8P7 SLC22A31 DPEP1 SNAI3 LOC100129697 ACSF3 LOC107984817 SPIRE2 PRDM7 LOC105371409 MIR4722 TCF25 PIEZO1 LOC100289580 CDK10 LOC101927793 AFG3L1P CDT1 TUBB3 MC1R SPATA2L MVD IL17C ZNF276 SNORD68 FAM157C SPG7 CPNE7 GALNS LOC101927817 GAS8 FANCA APRT CYBA ZNF778 PABPN1L LOC105371414 DEF8 TRAPPC2L CDH15 RPL13 VPS9D1-AS1 LOC105371419 CBFA2T3 CENPBD1 URAHP DBNDD1 SPATA33 VPS9D1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |