Assay Details
Target Gene Details
Entrez Gene ID: | 8312 |
Gene Name: | axin 1 |
Gene Aliases: |
AXIN, PPP1R49 |
Location: |
Chr.16:287440-355226 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AXIN1 | NM_003502.3 | NP_003493.1 | ||
NM_181050.2 | NP_851393.1 | |||
NR_134879.1 | ||||
XM_011522682.2 | XP_011520984.1 | |||
XM_011522683.2 | XP_011520985.1 | |||
XM_011522684.2 | XP_011520986.1 | |||
XM_011522686.1 | XP_011520988.1 | |||
XM_017023743.1 | XP_016879232.1 | |||
XM_017023744.1 | XP_016879233.1 | |||
XM_017023745.1 | XP_016879234.1 | |||
XM_017023746.1 | XP_016879235.1 | |||
XM_017023747.1 | XP_016879236.1 | |||
XM_017023748.1 | XP_016879237.1 | |||
AB208876.1 | BAD92113.1 | |||
AF009674.1 | AAC51624.1 | |||
BC044648.1 | AAH44648.1 | |||
KF615847.1 | ||||
KF615848.1 | ||||
KF615849.1 | ||||
KF615850.1 | ||||
KF615851.1 | ||||
KF615852.1 | ||||
KF615853.1 | ||||
KF615854.1 | ||||
KU178413.1 | ||||
KU178414.1 | ||||
KU178415.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3637568 | Chr.16:300040 - 319624 on Build GRCh38 | Gain | AXIN1 |
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | HBA1 METTL26 LMF1 ABCA17P PDPK1 RPL23AP5 RGS11 PGP MLST8 MEIOB GNG13 LOC100287175 BRICD5 MIR6768 NARFL PRR25 WDR90 RAB40C C16orf91 CCDC78 SNORD60 MIR662 ATP6V0C LOC100134368 CCNF CASKIN1 JMJD8 TPSG1 SSTR5 PRSS27 CHTF18 MIR6859-4 TSC2 CEMP1 MIR3177 RHOT2 MRPS34 LINC00235 LOC652276 CLCN7 ERVK13-1 POLR3K MIR3178 LOC729652 NTN3 TSR3 MIR3677 TPSD1 SOX8 DECR2 RPL3L PRR35 SPSB3 ARHGDIG LOC105371045 TPSB2 NTHL1 STUB1 EME2 SSTR5-AS1 IFT140 DDX11L10 RHBDL1 BAIAP3 GFER MIR4516 GNPTG SNHG19 PTX4 SNRNP25 AMDHD2 RAB11FIP3 NPRL3 LOC105371049 RPUSD1 TPSAB1 ABCA3 PKD1 HBZ SNORA10 LOC105371038 CACNA1H WASIR2 WFIKKN1 C1QTNF8 LUC7L PIGQ LOC107987233 HS3ST6 PDIA2 SLC9A3R2 UNKL LOC107984876 MIR6767 KCTD5 PRSS29P NME3 LOC106660606 TRAF7 CCDC154 MCRIP2 TMEM204 NHLRC4 ECI1 HBQ1 FLJ42627 FBXL16 ZNF598 TELO2 C16orf59 SNHG9 IGFALS MAPK8IP3 RPS2 SNORA64 NME4 AXIN1 NDUFB10 TMEM8A NOXO1 MSRB1 WDR24 HN1L MIR3180-5 FAHD1 FAM234A LOC105371184 LMF1-AS1 LOC105371046 NUBP2 LOC105371047 MPG MSLN TBC1D24 TBL3 UBE2I MIR3176 LOC101929280 MIR1225 FAM173A NPW HBA2 HAGH RNPS1 LINC00254 METRN MRPL28 HBM MIR6511B1 RNF151 MIR940 CRAMP1 HAGHL CAPN15 SNORA78 SYNGR3 E4F1 RHBDF1 DNASE1L2 MIR4717 MIR5587 RAB26 |
esv3637564 | Chr.16:287941 - 370218 on Build GRCh38 | Gain | MRPL28 AXIN1 |
nsv952896 | Chr.16:273501 - 321500 on Build GRCh38 | Deletion | PDIA2 RGS11 AXIN1 ARHGDIG |
esv3637563 | Chr.16:263248 - 505590 on Build GRCh38 | Gain | TMEM8A FAM234A RPL23AP5 MRPL28 DECR2 RAB11FIP3 PDIA2 RGS11 NME4 AXIN1 ARHGDIG LOC100134368 |
nsv570951 | Chr.16:279671 - 322597 on Build GRCh38 | Loss | PDIA2 AXIN1 ARHGDIG |
dgv32n68 | Chr.16:245507 - 476226 on Build GRCh38 | Loss | TMEM8A FAM234A RPL23AP5 MRPL28 DECR2 RAB11FIP3 PDIA2 RGS11 NME4 AXIN1 ARHGDIG LOC100134368 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |