Assay Details
Target Gene Details
Entrez Gene ID: | 9727 |
Gene Name: | RAB11 family interacting protein 3 |
Gene Aliases: |
CART1, Rab11-FIP3 |
Location: |
Chr.16:425621-522485 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RAB11FIP3 | NM_014700.3 | NP_055515.1 | ||
XM_005255713.3 | XP_005255770.1 | |||
XM_005255717.3 | XP_005255774.1 | |||
XM_005255718.3 | XP_005255775.1 | |||
XM_017023907.1 | XP_016879396.1 | |||
AB014565.1 | ||||
AF395731.1 | AAL12940.1 | |||
BC051360.1 | AAH51360.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | WFIKKN1 AMDHD2 SLC9A3R2 RHBDL1 RGS11 HBA2 MIR3677 NARFL MLST8 NOXO1 METTL26 UBE2I BAIAP3 HAGHL MPG LINC00235 RPL3L MIR3178 TPSD1 NPW MEIOB TBL3 HN1L FAM234A CAPN15 NME3 NTN3 NME4 C1QTNF8 SNHG9 C16orf59 SYNGR3 C16orf91 ABCA3 SNRNP25 TPSB2 RNF151 JMJD8 MIR6767 HBQ1 RPS2 LOC107984876 ZNF598 GNPTG SNORA64 NUBP2 ATP6V0C PRSS29P ERVK13-1 LOC107987233 HS3ST6 AXIN1 LOC729652 PRR25 LUC7L PDIA2 SSTR5 LOC105371049 TSR3 CCNF NTHL1 RHBDF1 E4F1 MCRIP2 RAB40C FAM173A MSRB1 GNG13 LOC100134368 NHLRC4 MIR6511B1 CEMP1 PRR35 TMEM8A RNPS1 PKD1 METRN LOC105371184 MIR662 MSLN WASIR2 UNKL PGP LOC105371038 MIR940 LINC00254 BRICD5 NPRL3 SNORD60 TELO2 MIR6768 SPSB3 CCDC78 KCTD5 TPSG1 MIR3176 SNORA78 ABCA17P FLJ42627 LOC101929280 LOC105371045 NDUFB10 IGFALS TMEM204 PIGQ HBM STUB1 TSC2 WDR24 DNASE1L2 PDPK1 DDX11L10 MIR1225 DECR2 POLR3K CRAMP1 CACNA1H MIR5587 RPL23AP5 GFER HBA1 MIR4516 SSTR5-AS1 CASKIN1 IFT140 TPSAB1 LOC106660606 WDR90 LOC105371047 LOC100287175 TBC1D24 MIR3180-5 LOC652276 LMF1 MIR3177 SNORA10 CLCN7 MIR4717 RPUSD1 TRAF7 HBZ MAPK8IP3 CCDC154 SOX8 FAHD1 LOC105371046 EME2 ARHGDIG SNHG19 PRSS27 ECI1 LMF1-AS1 CHTF18 MIR6859-4 RAB11FIP3 MRPS34 HAGH RHOT2 RAB26 MRPL28 FBXL16 PTX4 |
esv3637576 | Chr.16:439034 - 480097 on Build GRCh38 | Gain | RAB11FIP3 |
esv3637563 | Chr.16:263248 - 505590 on Build GRCh38 | Gain | RGS11 AXIN1 MRPL28 ARHGDIG LOC100134368 PDIA2 DECR2 FAM234A RPL23AP5 NME4 RAB11FIP3 TMEM8A |
nsv526903 | Chr.16:451362 - 810673 on Build GRCh38 | Loss | LOC107987233 WFIKKN1 PRR25 WDR90 CCDC78 MIR3176 LOC100287175 MCRIP2 RAB40C FAM173A RHBDL1 LOC101929280 RPUSD1 GNG13 NARFL NHLRC4 METTL26 PIGQ PRR35 HAGHL STUB1 WDR24 LINC00235 CAPN15 CHTF18 METRN LOC105371184 RAB11FIP3 MIR662 MSLN JMJD8 RHOT2 MIR5587 LOC105371038 FBXL16 |
nsv570954 | Chr.16:342461 - 530124 on Build GRCh38 | Loss | LINC00235 AXIN1 MRPL28 LOC100134368 DECR2 CAPN15 RPL23AP5 NME4 RAB11FIP3 TMEM8A |
dgv32n68 | Chr.16:245507 - 476226 on Build GRCh38 | Loss | RGS11 AXIN1 MRPL28 ARHGDIG LOC100134368 PDIA2 DECR2 FAM234A RPL23AP5 NME4 RAB11FIP3 TMEM8A |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |