Assay Details
Target Gene Details
Entrez Gene ID: | 1428 |
Gene Name: | crystallin mu |
Gene Aliases: |
DFNA40, THBP |
Location: |
Chr.16:21258518-21303136 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CRYM | NM_001888.4 | NP_001879.1 | ||
BG033512.1 | ||||
BI860684.1 | ||||
BX648477.1 | CAI46030.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv34235 | Chr.16:21276264 - 21986230 on Build GRCh38 | Gain | UQCRC2 OTOA IGSF6 CRYM-AS1 CRYM NPIPB4 NPIPB3 RRN3P1 PDZD9 LOC101927814 SMG1P3 SNX29P1 LOC100190986 METTL9 MIR3680-1 SLC7A5P2 |
nsv571623 | Chr.16:21084049 - 21341186 on Build GRCh38 | Gain | ANKS4B DNAH3 ZP2 CRYM-AS1 CRYM TMEM159 |
esv3638164 | Chr.16:21185596 - 21338776 on Build GRCh38 | Gain | ANKS4B ZP2 CRYM-AS1 CRYM |
nsv436835 | Chr.16:18930652 - 21500976 on Build GRCh38 | Insertion | UMOD GP2 ACSM5 ZP2 SYT17 ITPRIPL2 NPIPB3 GPR139 TMC5 SNX29P1 DNAH3 LYRM1 ERI2 CLEC19A KNOP1 ACSM2A LOC81691 ANKS4B DCUN1D3 GPRC5B CCP110 ACSM3 TMC7 CRYM-AS1 ACSM2B CRYM COQ7 THUMPD1 SMG1P3 PDILT IQCK LOC102723385 LOC100190986 ACSM1 GDE1 TMEM159 C16orf62 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |