Assay Details
Target Gene Details
Entrez Gene ID: | 8399 |
Gene Name: | phospholipase A2 group X |
Gene Aliases: |
GXPLA2, GXSPLA2, SPLA2, sPLA2-X |
Location: |
Chr.16:14672548-14694663 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PLA2G10 | NM_003561.2 | NP_003552.1 | ||
NR_133651.1 | ||||
XM_011522695.2 | XP_011520997.1 | |||
BC069539.1 | AAH69539.1 | |||
BC106731.1 | AAI06732.1 | |||
BC106732.1 | AAI06733.1 | |||
BC111804.1 | ||||
BM739473.1 | ||||
CR456885.1 | CAG33166.1 | |||
U95301.1 | AAB64410.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833144 | Chr.16:14587299 - 14813650 on Build GRCh38 | Gain | LOC100652777 NPIPA3 PLA2G10 LOC105371095 NPIPA2 BFAR PARN LOC105376747 |
dgv2738n100 | Chr.16:14617525 - 15031584 on Build GRCh38 | Gain | MIR1972-1 LOC100652777 MIR3180-1 ABCC6P2 NPIPA3 PLA2G10 LOC105371095 LOC728138 MIR6511A1 BFAR PDXDC1 MIR6770-1 MIR3179-1 LOC100288162 NPIPA1 NPIPA2 NOMO1 MIR3670-1 PARN LOC105376747 |
nsv984262 | Chr.16:14678216 - 14774331 on Build GRCh38 | Duplication | LOC100652777 NPIPA3 PLA2G10 LOC105371095 NPIPA2 LOC105376747 |
esv2758632 | Chr.16:14528198 - 15272693 on Build GRCh38 | Gain+Loss | MIR1972-1 LOC100652777 MIR3180-1 ABCC6P2 NPIPA3 PLA2G10 MIR6511A1 BFAR PDXDC1 MIR3179-1 NPIPA1 MIR3180-4 PKD1P6-NPIPP1 MIR6511B2 PARN LOC105376747 NTAN1 LOC105371095 LOC728138 RRN3 MIR6770-1 LOC100505915 LOC100288162 NPIPA2 NOMO1 MIR3670-1 |
nsv1740 | Chr.16:14654306 - 14769738 on Build GRCh38 | Deletion | LOC100652777 NPIPA3 PLA2G10 LOC105371095 NPIPA2 BFAR LOC105376747 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |