Assay Details
Target Gene Details
Entrez Gene ID: | 4913 |
Gene Name: | nth-like DNA glycosylase 1 |
Gene Aliases: |
FAP3, NTH1, OCTS3, hNTH1 |
Location: |
Chr.16:2039815-2047872 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NTHL1 | NM_001318193.1 | NP_001305122.1 | ||
NM_001318194.1 | NP_001305123.1 | |||
NM_002528.6 | NP_002519.1 | |||
XM_017023253.1 | XP_016878742.1 | |||
AB001575.1 | BAA19413.1 | |||
BC000391.2 | AAH00391.2 | |||
BC003014.1 | AAH03014.1 | |||
BE900738.1 | ||||
BG468617.1 | ||||
U79718.1 | AAB41534.1 | |||
U81285.1 | AAC51136.1 | |||
Y09687.1 | CAA70865.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | MRPL28 TBL3 SNORA10 WDR90 DECR2 ABCA3 LINC00235 HAGH LOC105371049 NARFL NME3 MCRIP2 TSR3 HBZ LINC00254 PGP SNHG19 POLR3K LOC100134368 UNKL MIR4516 BRICD5 HN1L WDR24 HBA1 CEMP1 CCNF TPSAB1 FAM173A LOC105371047 ARHGDIG ABCA17P RNF151 C16orf91 IGFALS HS3ST6 LOC100287175 HBQ1 MIR1225 PIGQ TPSG1 MIR6859-4 RPL23AP5 LOC105371046 MRPS34 PRSS29P CCDC154 SNHG9 TMEM8A ECI1 LMF1 RHOT2 DDX11L10 MIR6767 RHBDF1 SNORA64 GNPTG RAB11FIP3 MIR3176 E4F1 WASIR2 TRAF7 UBE2I CAPN15 MSRB1 RPUSD1 SNRNP25 JMJD8 NPRL3 TSC2 LOC107987233 RNPS1 ERVK13-1 LOC105371038 CHTF18 RAB40C MSLN LOC107984876 NME4 FAHD1 MLST8 NOXO1 NDUFB10 HBA2 MIR4717 LOC101929280 LOC105371184 TPSB2 SNORA78 METTL26 HAGHL RPL3L MIR3677 NUBP2 WFIKKN1 MIR6511B1 NTHL1 C16orf59 RAB26 LOC729652 SPSB3 CASKIN1 IFT140 FAM234A NPW NHLRC4 SOX8 PDPK1 LOC106660606 CRAMP1 SNORD60 SLC9A3R2 MIR3180-5 PRR35 KCTD5 METRN AMDHD2 RHBDL1 TELO2 SYNGR3 PTX4 MPG RGS11 LOC652276 LOC105371045 MIR5587 AXIN1 LUC7L LMF1-AS1 TPSD1 BAIAP3 ATP6V0C GNG13 CACNA1H MIR940 DNASE1L2 STUB1 MIR3177 C1QTNF8 HBM PRSS27 MIR3178 TBC1D24 NTN3 CLCN7 MEIOB MIR6768 PKD1 ZNF598 FBXL16 SSTR5-AS1 SSTR5 RPS2 CCDC78 TMEM204 GFER PDIA2 FLJ42627 MAPK8IP3 MIR662 PRR25 EME2 |
nsv471072 | Chr.16:1971892 - 2317508 on Build GRCh38 | Loss | SYNGR3 TBL3 MLST8 NOXO1 MIR4717 ECI1 ABCA3 LOC105371049 MIR3677 MIR940 PGP DNASE1L2 SNHG19 MIR6511B1 NTHL1 MIR4516 BRICD5 E4F1 RAB26 CASKIN1 TRAF7 NPW PKD1 ZNF598 LOC106660606 SNORD60 SLC9A3R2 MIR3180-5 MIR1225 TSC2 GFER RNPS1 |
nsv952901 | Chr.16:1888800 - 2135899 on Build GRCh38 | Deletion | SYNGR3 TBL3 SNORA10 NOXO1 NDUFB10 SNHG9 NPW PKD1 ZNF598 MSRB1 LOC105371049 SNORA78 RNF151 SLC9A3R2 RPL3L RPS2 HS3ST6 MIR1225 TSC2 GFER MIR6511B1 NTHL1 MIR4516 SNORA64 |
nsv1160329 | Chr.16:1961727 - 2128574 on Build GRCh38 | Deletion | SYNGR3 TBL3 SNORA10 NOXO1 NDUFB10 SNHG9 NPW PKD1 ZNF598 LOC105371049 SNORA78 RNF151 SLC9A3R2 RPS2 MIR1225 TSC2 GFER MIR6511B1 NTHL1 SNORA64 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |