Assay Details
Target Gene Details
Entrez Gene ID: | 9717 |
Gene Name: | SEC14 like lipid binding 5 |
Gene Aliases: |
PRELID4B |
Location: |
Chr.16:4958317-5019155 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SEC14L5 | NM_014692.1 | 1 | 84 | NP_055507.1 |
AB007880.2 | 1 | 84 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3637714 | Chr.16:4958035 - 5079276 on Build GRCh38 | Gain | C16orf89 SEC14L5 ALG1 NAGPA NAGPA-AS1 |
esv3637713 | Chr.16:4957958 - 5307305 on Build GRCh38 | Gain | RBFOX1 C16orf89 SEC14L5 ALG1 NAGPA NAGPA-AS1 EEF2KMT |
esv25186 | Chr.16:4858500 - 5142800 on Build GRCh38 | Gain | UBN1 C16orf89 SEC14L5 PPL ALG1 NAGPA NAGPA-AS1 EEF2KMT |
esv2761893 | Chr.16:4941276 - 4995458 on Build GRCh38 | Loss | SEC14L5 |
esv2422516 | Chr.16:4775920 - 5160979 on Build GRCh38 | Deletion | GLYR1 UBN1 C16orf89 SEPT12 SEC14L5 PPL SMIM22 ROGDI ALG1 NAGPA NAGPA-AS1 EEF2KMT |
nsv1046986 | Chr.16:4049000 - 5555721 on Build GRCh38 | Gain | DNAJA3 GLYR1 SMIM22 ANKS3 SRL NAGPA NMRAL1 EEF2KMT PAM16 MGRN1 SEPT12 MIR6769A UBALD1 ROGDI ZNF500 TFAP4 CORO7 C16orf96 LINC01569 GLIS2-AS1 RBFOX1 C16orf89 PPL GLIS2 ALG1 VASN NAGPA-AS1 NUDT16L1 ADCY9 C16orf71 LOC105371063 UBN1 SEC14L5 HMOX2 CDIP1 CORO7-PAM16 |
nsv528586 | Chr.16:4951303 - 5021669 on Build GRCh38 | Gain | SEC14L5 |
esv2760069 | Chr.16:4922849 - 5374828 on Build GRCh38 | Gain+Loss | RBFOX1 C16orf89 SEC14L5 PPL ALG1 NAGPA NAGPA-AS1 EEF2KMT |
nsv1039730 | Chr.16:4902193 - 4970782 on Build GRCh38 | Loss | SEC14L5 PPL |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |