Assay Details
Target Gene Details
Entrez Gene ID: | 1632 |
Gene Name: | enoyl-CoA delta isomerase 1 |
Gene Aliases: |
DCI |
Location: |
Chr.16:2239872-2251601 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ECI1 | NM_001178029.1 | NP_001171500.1 | ||
NM_001919.3 | NP_001910.2 | |||
AK291127.1 | ||||
BC000762.2 | AAH00762.1 | |||
BC002746.2 | AAH02746.1 | |||
BC009631.1 | AAH09631.1 | |||
BC019316.1 | AAH19316.1 | |||
BC020228.1 | AAH20228.1 | |||
BQ773923.1 | ||||
L24774.1 | AAA35485.1 | |||
Z25820.1 | CAA81065.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv571200 | Chr.16:2209190 - 2244889 on Build GRCh38 | Loss | E4F1 ECI1 MLST8 BRICD5 DNASE1L2 PGP |
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | HBQ1 MSRB1 PDPK1 IGFALS TPSB2 WFIKKN1 MSLN NPRL3 HBA2 LOC100134368 CCDC154 KCTD5 TPSG1 TMEM204 METTL26 SSTR5 MIR662 METRN SNORD60 WDR90 RNPS1 ABCA17P CACNA1H MLST8 MPG RPL3L HN1L ERVK13-1 PGP PDIA2 MIR3177 MIR3178 WDR24 UNKL TMEM8A MIR5587 NUBP2 DECR2 PTX4 RPS2 ECI1 C1QTNF8 NTHL1 EME2 LOC729652 PIGQ SNORA78 MAPK8IP3 WASIR2 RAB40C SOX8 LOC105371047 LINC00254 MIR6859-4 TBC1D24 HS3ST6 BAIAP3 FLJ42627 CCNF LOC105371184 SPSB3 MIR3176 FAM234A MCRIP2 CHTF18 HBZ LOC106660606 LOC101929280 RPL23AP5 SNRNP25 ARHGDIG GFER CCDC78 LUC7L LINC00235 E4F1 MIR6511B1 FBXL16 PRSS29P NDUFB10 NHLRC4 PKD1 MIR1225 LOC105371045 LMF1-AS1 DDX11L10 SLC9A3R2 MEIOB CEMP1 MIR4516 TPSD1 ATP6V0C C16orf59 LOC107987233 LOC105371038 MIR3180-5 SNHG9 JMJD8 PRR35 RNF151 POLR3K PRSS27 FAM173A LOC100287175 RAB26 HBA1 NARFL SNORA10 UBE2I FAHD1 MRPL28 MIR940 SYNGR3 RHBDF1 RHBDL1 AXIN1 NPW GNPTG HAGHL RGS11 CRAMP1 RPUSD1 IFT140 BRICD5 LOC105371049 AMDHD2 STUB1 MIR6768 SSTR5-AS1 PRR25 ABCA3 CLCN7 GNG13 DNASE1L2 C16orf91 MIR6767 SNHG19 NME3 CASKIN1 NOXO1 NTN3 LOC107984876 ZNF598 MRPS34 TBL3 TPSAB1 HAGH RHOT2 RAB11FIP3 LOC105371046 TELO2 SNORA64 MIR4717 LMF1 TSR3 NME4 TSC2 HBM TRAF7 LOC652276 CAPN15 MIR3677 |
nsv471072 | Chr.16:1971892 - 2317508 on Build GRCh38 | Loss | E4F1 ECI1 MIR6511B1 NTHL1 BRICD5 LOC105371049 PKD1 MIR1225 ABCA3 SLC9A3R2 MIR4516 SNORD60 DNASE1L2 SNHG19 RNPS1 MIR3180-5 CASKIN1 NOXO1 ZNF598 MLST8 TBL3 PGP RAB26 LOC106660606 MIR4717 TSC2 MIR940 SYNGR3 NPW GFER TRAF7 MIR3677 |
dgv4861n54 | Chr.16:2083900 - 2246850 on Build GRCh38 | Loss | MIR3180-5 E4F1 ECI1 CASKIN1 MIR6511B1 MLST8 BRICD5 LOC105371049 PKD1 MIR1225 PGP RAB26 TSC2 MIR4516 TRAF7 SNORD60 DNASE1L2 SNHG19 |
dgv4863n54 | Chr.16:2141733 - 2274302 on Build GRCh38 | Loss | E4F1 ECI1 CASKIN1 MLST8 BRICD5 PGP RAB26 LOC106660606 MIR940 TRAF7 SNORD60 DNASE1L2 SNHG19 RNPS1 MIR3677 |
nsv457351 | Chr.16:2156590 - 2274302 on Build GRCh38 | Loss | LOC106660606 E4F1 ECI1 CASKIN1 MIR940 MLST8 BRICD5 TRAF7 DNASE1L2 PGP RNPS1 MIR3677 |
nsv1052260 | Chr.16:2074546 - 2629655 on Build GRCh38 | Gain | E4F1 ECI1 MIR6511B1 BRICD5 LOC105371049 AMDHD2 PKD1 PDPK1 MIR1225 LOC729652 MIR6768 ABCA3 CEMP1 MIR4516 SNORD60 DNASE1L2 ATP6V0C MIR6767 TBC1D24 SNHG19 C16orf59 RNPS1 MIR3180-5 ABCA17P CCNF CASKIN1 NTN3 MLST8 PGP RAB26 LOC106660606 MIR4717 MIR3178 TSC2 MIR940 TRAF7 LOC652276 MIR3677 |
nsv952902 | Chr.16:2143400 - 2339999 on Build GRCh38 | Deletion | E4F1 ECI1 CASKIN1 MLST8 BRICD5 PGP RAB26 LOC106660606 MIR4717 ABCA3 MIR940 TRAF7 SNORD60 DNASE1L2 SNHG19 RNPS1 MIR3677 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |