Assay Details
Target Gene Details
Entrez Gene ID: | 3029 |
Gene Name: | hydroxyacylglutathione hydrolase |
Gene Aliases: |
GLO2, GLX2, GLXII, HAGH1 |
Location: |
Chr.16:1809099-1827194 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HAGH | NM_001040427.1 | NP_001035517.1 | ||
NM_001286249.1 | NP_001273178.1 | |||
NM_005326.4 | NP_005317.2 | |||
XM_011522469.2 | XP_011520771.1 | |||
XM_011522470.2 | XP_011520772.1 | |||
XM_017023195.1 | XP_016878684.1 | |||
AK290155.1 | ||||
AK298174.1 | ||||
AK299173.1 | ||||
AK299993.1 | ||||
AK308825.1 | ||||
BC000840.1 | AAH00840.1 | |||
BC002627.2 | ||||
BI758286.1 | ||||
BU176541.1 | ||||
DB502986.1 | ||||
X90999.1 | CAA62483.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv517196 | Chr.16:1729694 - 1844960 on Build GRCh38 | Gain+Loss | MRPS34 MAPK8IP3 FAHD1 NME3 MIR3177 SPSB3 NUBP2 EME2 MEIOB IGFALS HAGH |
nsv571175 | Chr.16:1806439 - 1875278 on Build GRCh38 | Loss | FAHD1 MEIOB HAGH |
nsv1040440 | Chr.16:1811739 - 1866102 on Build GRCh38 | Gain | FAHD1 MEIOB HAGH |
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | MIR3177 SNORA64 SNHG9 CLCN7 LMF1 LOC105371045 SLC9A3R2 BRICD5 IGFALS UNKL LOC105371038 CASKIN1 PRR35 LOC105371047 STUB1 ATP6V0C CRAMP1 ZNF598 LOC729652 SSTR5-AS1 SNORD60 MAPK8IP3 PRR25 LUC7L WASIR2 RAB26 HBQ1 UBE2I MIR6768 LOC101929280 RPUSD1 MEIOB ERVK13-1 SNORA78 MIR6511B1 MRPS34 C16orf59 WDR90 ECI1 FBXL16 RHBDF1 C1QTNF8 MIR662 RHOT2 NTN3 ABCA3 TMEM204 CEMP1 BAIAP3 MIR3180-5 SNRNP25 SNHG19 FAM173A LOC107984876 TPSB2 IFT140 SSTR5 MSRB1 TPSG1 HBA1 MIR6859-4 SYNGR3 DDX11L10 RPL23AP5 AMDHD2 MIR3176 MLST8 HBA2 HAGHL RGS11 NPRL3 HAGH GNG13 TBC1D24 RAB11FIP3 WDR24 LMF1-AS1 PRSS29P MIR6767 TMEM8A NME3 LOC100287175 LOC652276 ARHGDIG SOX8 CHTF18 MCRIP2 PGP FLJ42627 LINC00254 HBZ MIR3677 JMJD8 NPW TSC2 E4F1 LINC00235 LOC105371184 EME2 GFER FAHD1 DECR2 CCDC154 MIR4717 METRN POLR3K MIR940 MIR1225 SPSB3 METTL26 HS3ST6 MPG LOC106660606 KCTD5 TBL3 LOC100134368 WFIKKN1 RPL3L MIR4516 MSLN RNPS1 CCNF PDIA2 CAPN15 LOC107987233 NTHL1 PTX4 HBM DNASE1L2 NME4 NHLRC4 HN1L RAB40C RPS2 SNORA10 RNF151 TSR3 NUBP2 TPSD1 MRPL28 LOC105371046 RHBDL1 NARFL LOC105371049 TPSAB1 FAM234A MIR5587 CCDC78 MIR3178 PRSS27 PKD1 PDPK1 TELO2 TRAF7 NDUFB10 PIGQ GNPTG ABCA17P CACNA1H AXIN1 C16orf91 NOXO1 |
nsv1051692 | Chr.16:1505070 - 1866102 on Build GRCh38 | Loss | MAPK8IP3 MIR3177 SPSB3 HN1L NUBP2 EME2 IFT140 MEIOB LOC105371046 IGFALS MRPS34 FAHD1 NME3 LOC105371047 CRAMP1 TMEM204 TELO2 HAGH |
nsv1042986 | Chr.16:1784723 - 1866102 on Build GRCh38 | Loss | FAHD1 NUBP2 MEIOB IGFALS HAGH |
nsv518641 | Chr.16:1645775 - 1835631 on Build GRCh38 | Loss | MAPK8IP3 MIR3177 SPSB3 HN1L NUBP2 EME2 MEIOB IGFALS MRPS34 FAHD1 NME3 CRAMP1 HAGH |
nsv457337 | Chr.16:1813519 - 1855005 on Build GRCh38 | Loss | FAHD1 MEIOB HAGH |
nsv952900 | Chr.16:1724500 - 1827999 on Build GRCh38 | Deletion | MRPS34 MAPK8IP3 FAHD1 NME3 MIR3177 SPSB3 NUBP2 EME2 IGFALS HAGH |
nsv571177 | Chr.16:1817039 - 1897015 on Build GRCh38 | Gain | LINC00254 FAHD1 MEIOB HAGH |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |