Assay Details
Target Gene Details
Entrez Gene ID: | 64223 |
Gene Name: | MTOR associated protein, LST8 homolog |
Gene Aliases: |
GBL, GbetaL, LST8, POP3, WAT1 |
Location: |
Chr.16:2205177-2209417 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MLST8 | NM_001199173.1 | NP_001186102.1 | ||
NM_001199175.1 | NP_001186104.1 | |||
NM_022372.4 | NP_071767.3 | |||
XM_005255478.2 | XP_005255535.1 | |||
XM_005255479.2 | XP_005255536.1 | |||
XM_017023548.1 | XP_016879037.1 | |||
XM_017023549.1 | XP_016879038.1 | |||
XM_017023550.1 | 1 | 128 | XP_016879039.1 | |
XM_017023551.1 | XP_016879040.1 | |||
XM_017023552.1 | 1 | 134 | XP_016879041.1 | |
AK021536.1 | ||||
AK098762.1 | ||||
AK302201.1 | ||||
AK304390.1 | 1 | 221 | ||
AU117583.1 | ||||
AY223837.1 | AAO73410.1 | |||
BC001313.2 | AAH01313.1 | |||
BC052292.1 | 1 | 200 | AAH52292.1 | |
BC088354.1 | AAH88354.1 | |||
BI761765.1 | ||||
DA382215.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | HS3ST6 LOC105371049 SNHG9 LINC00254 DDX11L10 LOC100134368 CLCN7 EME2 FAM234A CHTF18 NTHL1 FLJ42627 RPS2 MSLN TPSD1 LOC652276 PTX4 CASKIN1 RAB11FIP3 MRPS34 NPRL3 JMJD8 MIR6511B1 ECI1 NTN3 TPSAB1 MIR6768 IFT140 ERVK13-1 TSC2 NME4 CRAMP1 SSTR5 RPL3L MRPL28 PKD1 HBM STUB1 LOC107987233 MIR3176 WFIKKN1 CACNA1H NOXO1 BAIAP3 TMEM8A AXIN1 C16orf59 ARHGDIG GNG13 WASIR2 PRR35 LUC7L ABCA3 C16orf91 LOC105371047 MIR4516 MIR3180-5 SNORA10 RPUSD1 WDR90 SNORA78 LOC105371038 MIR3178 GNPTG TPSG1 IGFALS RNF151 LOC100287175 ABCA17P NARFL SNORA64 PIGQ LOC101929280 HAGH FBXL16 CCNF KCTD5 UBE2I E4F1 ZNF598 AMDHD2 FAM173A LOC105371046 TRAF7 MIR1225 MIR5587 MCRIP2 RAB26 TPSB2 MPG MIR662 NPW MEIOB CEMP1 LMF1 HBA2 SPSB3 RAB40C HAGHL HBZ C1QTNF8 PRR25 MIR4717 GFER SLC9A3R2 NDUFB10 FAHD1 RNPS1 LOC729652 WDR24 TSR3 ATP6V0C MIR6859-4 PGP DECR2 RHBDL1 HN1L PDPK1 CCDC154 NUBP2 RGS11 LOC105371045 MIR3177 POLR3K TELO2 MIR940 SSTR5-AS1 SNRNP25 DNASE1L2 LOC107984876 HBQ1 RHBDF1 PDIA2 LOC106660606 MIR6767 UNKL METRN CAPN15 METTL26 TMEM204 MSRB1 RPL23AP5 MIR3677 CCDC78 HBA1 NME3 LMF1-AS1 MLST8 TBL3 RHOT2 BRICD5 PRSS29P MAPK8IP3 PRSS27 SNHG19 TBC1D24 LINC00235 SNORD60 LOC105371184 SOX8 NHLRC4 SYNGR3 |
nsv471072 | Chr.16:1971892 - 2317508 on Build GRCh38 | Loss | NOXO1 ZNF598 LOC105371049 MIR940 TRAF7 MIR1225 DNASE1L2 RAB26 NTHL1 LOC106660606 NPW MIR3677 CASKIN1 ABCA3 MIR4516 MIR3180-5 MIR4717 MLST8 TBL3 GFER SLC9A3R2 MIR6511B1 RNPS1 BRICD5 ECI1 SNHG19 TSC2 PGP SNORD60 PKD1 SYNGR3 E4F1 |
dgv4861n54 | Chr.16:2083900 - 2246850 on Build GRCh38 | Loss | MLST8 MIR6511B1 LOC105371049 BRICD5 ECI1 TRAF7 MIR1225 DNASE1L2 RAB26 SNHG19 TSC2 PGP SNORD60 PKD1 CASKIN1 MIR4516 MIR3180-5 E4F1 |
dgv4863n54 | Chr.16:2141733 - 2274302 on Build GRCh38 | Loss | MLST8 MIR940 RNPS1 BRICD5 ECI1 TRAF7 DNASE1L2 RAB26 SNHG19 LOC106660606 PGP SNORD60 MIR3677 CASKIN1 E4F1 |
nsv1711 | Chr.16:2199093 - 2232650 on Build GRCh38 | Insertion | PGP MLST8 BRICD5 E4F1 |
nsv457351 | Chr.16:2156590 - 2274302 on Build GRCh38 | Loss | PGP MLST8 MIR940 RNPS1 BRICD5 ECI1 TRAF7 DNASE1L2 MIR3677 CASKIN1 LOC106660606 E4F1 |
nsv1052260 | Chr.16:2074546 - 2629655 on Build GRCh38 | Gain | AMDHD2 LOC105371049 MIR940 TRAF7 MIR1225 DNASE1L2 C16orf59 RAB26 LOC106660606 MIR6767 CEMP1 LOC652276 MIR3677 CASKIN1 ABCA3 MIR4516 MIR3180-5 MIR4717 MLST8 MIR6511B1 RNPS1 BRICD5 ECI1 NTN3 MIR3178 MIR6768 SNHG19 TBC1D24 TSC2 LOC729652 ABCA17P ATP6V0C PGP SNORD60 PKD1 PDPK1 CCNF E4F1 |
nsv952902 | Chr.16:2143400 - 2339999 on Build GRCh38 | Deletion | MLST8 MIR940 RNPS1 BRICD5 ECI1 TRAF7 DNASE1L2 RAB26 SNHG19 LOC106660606 PGP SNORD60 MIR3677 CASKIN1 ABCA3 MIR4717 E4F1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |