Assay Details
Target Gene Details
Entrez Gene ID: | 6477 |
Gene Name: | siah E3 ubiquitin protein ligase 1 |
Gene Aliases: |
SIAH1A |
Location: |
Chr.16:48360531-48448440 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SIAH1 | NM_003031.3 | NP_003022.3 | ||
XM_006721246.1 | XP_006721309.1 | |||
XM_011523279.1 | XP_011521581.1 | |||
AK225895.1 | ||||
BC035562.1 | AAH35562.1 | |||
BC044920.1 | AAH44920.1 | |||
U76247.1 | AAC51907.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2758645 | Chr.16:48295526 - 48445843 on Build GRCh38 | Gain | LONP2 LOC100507577 SIAH1 |
nsv952668 | Chr.16:48365790 - 48384189 on Build GRCh38 | Duplication | SIAH1 |
nsv833219 | Chr.16:48256684 - 48425390 on Build GRCh38 | Gain | LONP2 LOC100507577 SIAH1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |