Assay Details
Target Gene Details
Entrez Gene ID: | 4350 |
Gene Name: | N-methylpurine DNA glycosylase |
Gene Aliases: |
AAG, ADPG, APNG, CRA36.1, MDG, Mid1, PIG11, PIG16, anpg |
Location: |
Chr.16:77019-85851 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MPG | NM_001015052.2 | NP_001015052.1 | ||
NM_001015054.2 | NP_001015054.1 | |||
NM_002434.3 | NP_002425.2 | |||
AK308654.1 | ||||
AL520052.3 | ||||
AY258284.1 | AAP82229.1 | |||
BC014991.1 | AAH14991.1 | |||
BG823448.1 | ||||
BM796301.1 | ||||
L10752.1 | AAF77073.1 | |||
M71215.1 | AAA58369.1 | |||
M74905.1 | AAA58627.1 | |||
S51033.1 | AAB19537.1 | |||
X56528.1 | CAA39875.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3637537 | Chr.16:65681 - 100253 on Build GRCh38 | Gain |
![]() ![]() ![]() |
nsv518291 | Chr.16:47354 - 254515 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv2690n100 | Chr.16:10765 - 207261 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv32706 | Chr.16:79410 - 95539 on Build GRCh38 | Loss |
![]() ![]() |
esv2760065 | Chr.16:31052 - 143601 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)