Assay Details
Target Gene Details
Entrez Gene ID: | 23199 |
Gene Name: | Gse1 coiled-coil protein |
Gene Aliases: |
CRHSP24, KIAA0182 |
Location: |
Chr.16:85169512-85676206 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GSE1 | XM_005255859.4 | XP_005255916.3 | ||
XM_005255860.3 | XP_005255917.3 | |||
XM_005255861.4 | XP_005255918.3 | |||
XM_017023084.1 | XP_016878573.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833312 | Chr.16:85158268 - 85327826 on Build GRCh38 | Loss | GSE1 LINC00311 MIR5093 |
dgv3059n100 | Chr.16:85068826 - 85219999 on Build GRCh38 | Gain | GSE1 LOC400548 FAM92B KIAA0513 |
nsv483044 | Chr.16:84166395 - 87066394 on Build GRCh38 | Gain | LINC01081 FOXL1 LOC105371374 MIR6774 LOC101928474 FENDRR FLJ30679 LOC440390 GINS2 MTHFSD COTL1 LOC101928557 LINC00311 MIR1910 MIR5093 LOC102724344 ATP2C2 EMC8 FOXF1 C16orf74 TAF1C LOC146513 IRF8 FOXC2 LOC400548 FAM92B LOC101928614 FOXC2-AS1 ADAD2 ZDHHC7 KCNG4 COX4I1 TLDC1 GSE1 LINC00917 KLHL36 LOC105376777 DNAAF1 KIAA0513 USP10 WFDC1 LINC01082 CRISPLD2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |