Assay Details
Target Gene Details
Entrez Gene ID: | 23199 |
Gene Name: | Gse1 coiled-coil protein |
Gene Aliases: |
CRHSP24, KIAA0182 |
Location: |
Chr.16:85169512-85676206 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GSE1 | XM_005255859.4 | XP_005255916.3 | ||
XM_005255860.3 | XP_005255917.3 | |||
XM_005255861.4 | XP_005255918.3 | |||
XM_017023084.1 | XP_016878573.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833312 | Chr.16:85158268 - 85327826 on Build GRCh38 | Loss | MIR5093 LINC00311 GSE1 |
dgv3059n100 | Chr.16:85068826 - 85219999 on Build GRCh38 | Gain | KIAA0513 LOC400548 FAM92B GSE1 |
nsv483044 | Chr.16:84166395 - 87066394 on Build GRCh38 | Gain | LOC146513 KCNG4 TLDC1 EMC8 GINS2 ADAD2 KLHL36 MIR6774 CRISPLD2 LINC01082 IRF8 KIAA0513 COX4I1 FENDRR LOC101928474 LOC440390 FLJ30679 TAF1C FOXF1 LOC400548 FAM92B LOC105371374 FOXC2-AS1 LINC00311 C16orf74 LOC102724344 MTHFSD GSE1 ZDHHC7 LINC01081 LINC00917 COTL1 MIR5093 DNAAF1 USP10 FOXC2 MIR1910 ATP2C2 WFDC1 LOC105376777 LOC101928614 LOC101928557 FOXL1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |