Assay Details
Target Gene Details
Entrez Gene ID: | 23162 |
Gene Name: | mitogen-activated protein kinase 8 interacting protein 3 |
Gene Aliases: |
JIP-3, JIP3, JSAP1, SYD2, syd |
Location: |
Chr.16:1706183-1770317 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MAPK8IP3 | NM_001040439.1 | NP_001035529.1 | ||
NM_001318852.1 | NP_001305781.1 | |||
NM_015133.4 | NP_055948.2 | |||
XM_005255190.2 | XP_005255247.1 | |||
XM_006720869.2 | XP_006720932.1 | |||
XM_011522429.2 | XP_011520731.1 | |||
XM_011522430.2 | XP_011520732.1 | |||
XM_011522431.2 | XP_011520733.1 | |||
XM_011522432.2 | XP_011520734.1 | |||
XM_017023079.1 | XP_016878568.1 | |||
XM_017023080.1 | XP_016878569.1 | |||
AB028989.2 | ||||
AB621811.1 | ||||
AK024437.1 | BAB15727.1 | |||
BC137123.1 | ||||
BC137124.1 | ||||
BC144486.1 | ||||
BC150266.1 | ||||
BY795428.2 | ||||
CN405914.1 | ||||
CR749696.1 | CAH18476.1 | |||
DA135446.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv4856n54 | Chr.16:1600945 - 1813519 on Build GRCh38 | Loss | IFT140 MIR3177 HAGH NUBP2 MRPS34 NME3 CRAMP1 LOC105371046 HN1L IGFALS SPSB3 MAPK8IP3 EME2 |
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | CASKIN1 PKD1 TSR3 MRPL28 MPG NUBP2 SNHG19 RAB40C MIR940 MIR4516 LOC106660606 LOC105371046 CCDC154 LOC100134368 BRICD5 STUB1 LOC105371047 LUC7L KCTD5 SNHG9 ABCA17P TMEM8A LOC105371049 SSTR5 SOX8 MIR3677 RAB26 LOC105371184 RPUSD1 LOC107987233 E4F1 NME4 MIR3176 NOXO1 TMEM204 DNASE1L2 CRAMP1 LOC729652 NARFL HBZ FAM234A NPRL3 LINC00254 HBQ1 HAGHL SNORA78 NHLRC4 JMJD8 GNPTG FAM173A RGS11 LOC100287175 ECI1 MIR3180-5 TPSG1 RAB11FIP3 TSC2 MIR6768 DDX11L10 NTHL1 PRR25 TBL3 CHTF18 NDUFB10 METRN LOC652276 MSLN LINC00235 HBA2 RNF151 C1QTNF8 MSRB1 AXIN1 TELO2 IGFALS WFIKKN1 C16orf91 SLC9A3R2 TPSD1 CCNF ARHGDIG PRSS27 HAGH ATP6V0C TRAF7 PDIA2 MRPS34 NTN3 SYNGR3 FAHD1 MIR5587 TPSAB1 PGP HBM TPSB2 BAIAP3 LMF1-AS1 SNRNP25 C16orf59 TBC1D24 RPL23AP5 SNORA10 WASIR2 SSTR5-AS1 MCRIP2 RHBDF1 RPS2 HBA1 PIGQ CEMP1 ZNF598 UNKL POLR3K RPL3L LOC101929280 MAPK8IP3 CACNA1H GNG13 GFER MIR6859-4 RHOT2 WDR24 HS3ST6 AMDHD2 LOC105371045 WDR90 MEIOB NME3 MIR662 LMF1 PTX4 HN1L DECR2 EME2 CAPN15 METTL26 UBE2I MIR3178 FLJ42627 CLCN7 IFT140 PDPK1 LOC105371038 MIR4717 PRR35 MIR3177 NPW LOC107984876 RHBDL1 MIR1225 PRSS29P RNPS1 SNORD60 ABCA3 CCDC78 MLST8 MIR6767 SNORA64 SPSB3 FBXL16 MIR6511B1 ERVK13-1 |
nsv1051692 | Chr.16:1505070 - 1866102 on Build GRCh38 | Loss | IFT140 MIR3177 HAGH NUBP2 MRPS34 FAHD1 MEIOB NME3 TMEM204 CRAMP1 LOC105371046 TELO2 HN1L IGFALS LOC105371047 SPSB3 MAPK8IP3 EME2 |
esv3637623 | Chr.16:1712012 - 1723067 on Build GRCh38 | Loss | MAPK8IP3 |
nsv1160328 | Chr.16:1704619 - 1797867 on Build GRCh38 | Deletion | MIR3177 NUBP2 IGFALS SPSB3 MRPS34 MAPK8IP3 EME2 NME3 |
nsv518641 | Chr.16:1645775 - 1835631 on Build GRCh38 | Loss | MIR3177 HAGH NUBP2 MRPS34 FAHD1 MEIOB NME3 CRAMP1 HN1L IGFALS SPSB3 MAPK8IP3 EME2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |