Assay Details
Target Gene Details
Entrez Gene ID: | 9727 |
Gene Name: | RAB11 family interacting protein 3 |
Gene Aliases: |
CART1, Rab11-FIP3 |
Location: |
Chr.16:425621-522485 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RAB11FIP3 | NM_014700.3 | NP_055515.1 | ||
XM_005255713.3 | XP_005255770.1 | |||
XM_005255717.3 | XP_005255774.1 | |||
XM_005255718.3 | XP_005255775.1 | |||
XM_017023907.1 | XP_016879396.1 | |||
AB014565.1 | ||||
AF395731.1 | AAL12940.1 | |||
BC051360.1 | AAH51360.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv521610 | Chr.16:430096 - 445538 on Build GRCh38 | Gain | RAB11FIP3 |
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | SNHG19 HBA1 LMF1-AS1 LOC729652 TBC1D24 WDR24 PDPK1 WDR90 NTN3 KCTD5 PGP PKD1 PTX4 CRAMP1 MIR3177 E4F1 CCDC154 LOC100134368 RHBDL1 TSC2 PRR35 ECI1 NHLRC4 LOC105371045 HAGHL TPSD1 STUB1 MIR662 HAGH ATP6V0C DECR2 HN1L HBA2 MIR3180-5 FLJ42627 DDX11L10 NUBP2 LINC00235 TPSG1 UBE2I FAHD1 TRAF7 RGS11 PRSS27 EME2 MSRB1 RPL23AP5 CACNA1H HBM MCRIP2 SLC9A3R2 SYNGR3 MIR940 TBL3 NDUFB10 RPS2 HBQ1 MIR3176 CASKIN1 SPSB3 SNHG9 CLCN7 SSTR5 NOXO1 RHOT2 TPSAB1 WASIR2 LOC100287175 FAM234A MIR1225 PRSS29P SNORD60 MIR5587 ERVK13-1 CEMP1 ZNF598 C16orf91 SNORA64 AXIN1 ARHGDIG FBXL16 TMEM204 GNPTG GFER MRPS34 TMEM8A JMJD8 MPG NPW NPRL3 NME3 MIR6511B1 RNPS1 PIGQ CAPN15 RPUSD1 ABCA17P METRN LINC00254 GNG13 FAM173A NARFL DNASE1L2 LOC107987233 MRPL28 LOC105371047 AMDHD2 LOC107984876 SNRNP25 NME4 MIR4717 METTL26 TSR3 LOC105371038 SNORA10 HS3ST6 TPSB2 CCDC78 CCNF LUC7L MIR6767 TELO2 SOX8 PDIA2 MIR3178 SNORA78 C16orf59 IFT140 WFIKKN1 RAB40C UNKL LMF1 MSLN ABCA3 MIR6768 LOC101929280 MLST8 MEIOB NTHL1 SSTR5-AS1 BRICD5 LOC105371046 RAB26 PRR25 LOC105371184 BAIAP3 IGFALS LOC652276 MAPK8IP3 RHBDF1 MIR6859-4 C1QTNF8 RNF151 MIR4516 RPL3L HBZ MIR3677 POLR3K RAB11FIP3 LOC106660606 CHTF18 LOC105371049 |
dgv4810n54 | Chr.16:380011 - 445662 on Build GRCh38 | Loss | TMEM8A DECR2 NME4 LOC100134368 RPL23AP5 RAB11FIP3 |
esv3637563 | Chr.16:263248 - 505590 on Build GRCh38 | Gain | TMEM8A DECR2 NME4 LOC100134368 RPL23AP5 ARHGDIG FAM234A MRPL28 RAB11FIP3 RGS11 AXIN1 PDIA2 |
nsv570956 | Chr.16:376432 - 437728 on Build GRCh38 | Gain | TMEM8A DECR2 NME4 LOC100134368 RPL23AP5 RAB11FIP3 |
nsv570954 | Chr.16:342461 - 530124 on Build GRCh38 | Loss | TMEM8A CAPN15 DECR2 NME4 LOC100134368 RPL23AP5 LINC00235 MRPL28 RAB11FIP3 AXIN1 |
nsv457303 | Chr.16:383623 - 445662 on Build GRCh38 | Loss | DECR2 NME4 LOC100134368 RPL23AP5 RAB11FIP3 |
dgv4809n54 | Chr.16:322597 - 451362 on Build GRCh38 | Loss | TMEM8A DECR2 NME4 LOC100134368 RPL23AP5 MRPL28 RAB11FIP3 AXIN1 |
dgv32n68 | Chr.16:245507 - 476226 on Build GRCh38 | Loss | TMEM8A DECR2 NME4 LOC100134368 RPL23AP5 ARHGDIG FAM234A MRPL28 RAB11FIP3 RGS11 AXIN1 PDIA2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |