Assay Details
Target Gene Details
Entrez Gene ID: | 254528 |
Gene Name: | meiosis specific with OB domains |
Gene Aliases: |
C16orf73, gs129 |
Location: |
Chr.16:1833983-1872178 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MEIOB | NM_001163560.2 | NP_001157032.1 | ||
NM_152764.2 | NP_689977.2 | |||
BC029829.1 | AAH29829.1 | |||
CD300613.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833119 | Chr.16:1834308 - 1911484 on Build GRCh38 | Loss | HS3ST6 MEIOB FAHD1 LINC00254 |
nsv571175 | Chr.16:1806439 - 1875278 on Build GRCh38 | Loss | HAGH MEIOB FAHD1 |
nsv1040440 | Chr.16:1811739 - 1866102 on Build GRCh38 | Gain | HAGH MEIOB FAHD1 |
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | CAPN15 NUBP2 RPL3L ZNF598 CASKIN1 METRN DECR2 NME3 CCNF POLR3K UBE2I SNRNP25 LOC100287175 C1QTNF8 RAB40C WDR24 NTN3 BRICD5 MSRB1 CCDC154 PRSS27 NARFL SNORA64 RGS11 LOC729652 HAGHL LUC7L IFT140 CCDC78 MPG RPL23AP5 LOC106660606 LMF1 WASIR2 MIR3677 PDPK1 CEMP1 RHOT2 MAPK8IP3 SSTR5-AS1 ARHGDIG MIR940 HBM TBL3 MIR3177 SNORD60 MIR3176 TPSG1 HBA1 MIR3180-5 NHLRC4 LOC107987233 FAM234A STUB1 TPSD1 NPW TBC1D24 RPS2 TSR3 SNHG19 MCRIP2 ATP6V0C SYNGR3 TSC2 PRSS29P WDR90 TMEM204 MSLN GNG13 MRPL28 FAM173A FBXL16 HBZ LOC101929280 MIR5587 PRR35 METTL26 LOC105371049 MRPS34 CLCN7 LOC105371038 RPUSD1 BAIAP3 SSTR5 LOC105371045 HS3ST6 UNKL RHBDF1 ABCA17P RAB26 MLST8 PKD1 MEIOB FAHD1 LOC105371047 MIR6859-4 SOX8 TPSAB1 MIR1225 LOC105371184 SNORA78 MIR662 MIR6768 EME2 LINC00254 HBQ1 SNHG9 NME4 RNF151 NTHL1 MIR6511B1 RAB11FIP3 HAGH ERVK13-1 MIR3178 JMJD8 NPRL3 E4F1 GFER CACNA1H CHTF18 IGFALS MIR4516 ABCA3 TPSB2 TELO2 PIGQ LOC105371046 RHBDL1 AXIN1 SPSB3 HBA2 PTX4 RNPS1 LMF1-AS1 LOC107984876 LOC652276 PRR25 MIR6767 FLJ42627 TRAF7 NOXO1 NDUFB10 C16orf59 SNORA10 ECI1 DDX11L10 GNPTG HN1L DNASE1L2 AMDHD2 PGP LINC00235 PDIA2 WFIKKN1 SLC9A3R2 CRAMP1 C16orf91 KCTD5 MIR4717 TMEM8A LOC100134368 |
nsv1051692 | Chr.16:1505070 - 1866102 on Build GRCh38 | Loss | NUBP2 SPSB3 HN1L NME3 HAGH MEIOB FAHD1 MRPS34 MAPK8IP3 LOC105371047 IGFALS MIR3177 CRAMP1 TMEM204 TELO2 LOC105371046 EME2 IFT140 |
nsv1046474 | Chr.16:1846666 - 1873690 on Build GRCh38 | Loss | MEIOB |
nsv1042986 | Chr.16:1784723 - 1866102 on Build GRCh38 | Loss | NUBP2 IGFALS HAGH MEIOB FAHD1 |
nsv457337 | Chr.16:1813519 - 1855005 on Build GRCh38 | Loss | HAGH MEIOB FAHD1 |
nsv571177 | Chr.16:1817039 - 1897015 on Build GRCh38 | Gain | HAGH MEIOB FAHD1 LINC00254 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |