Assay Details
Target Gene Details
Entrez Gene ID: | 11214 |
Gene Name: | A-kinase anchoring protein 13 |
Gene Aliases: |
AKAP-13, AKAP-Lbc, ARHGEF13, BRX, HA-3, Ht31, LBC, PRKA13, PROTO-LB, PROTO-LBC, c-lbc, p47 |
Location: |
Chr.15:85380616-85749358 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AKAP13 | NM_006738.5 | NP_006729.4 | ||
NM_007200.4 | NP_009131.2 | |||
AB055890.1 | BAB62913.1 | |||
AF406992.1 | AAL11723.1 | |||
AK125331.1 | ||||
BC050312.1 | AAH50312.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv4692n54 | Chr.15:85235254 - 85600273 on Build GRCh38 | Gain | AKAP13 GOLGA6L3 MIR7706 |
esv2761699 | Chr.15:85236347 - 85569198 on Build GRCh38 | Gain | AKAP13 GOLGA6L3 MIR7706 |
esv3637111 | Chr.15:85291627 - 85464874 on Build GRCh38 | Gain | AKAP13 MIR7706 |
esv3637112 | Chr.15:85384885 - 85468079 on Build GRCh38 | Gain | AKAP13 |
dgv323n27 | Chr.15:85282336 - 85600273 on Build GRCh38 | Gain | AKAP13 MIR7706 |
dgv2647n100 | Chr.15:85268380 - 85470404 on Build GRCh38 | Gain | AKAP13 MIR7706 |
nsv518064 | Chr.15:85178426 - 85605928 on Build GRCh38 | Gain | AKAP13 LOC642423 LOC101929479 GOLGA6L3 MIR7706 |
dgv4694n54 | Chr.15:85344022 - 85600273 on Build GRCh38 | Gain | AKAP13 MIR7706 |
dgv754e212 | Chr.15:85181918 - 85597222 on Build GRCh38 | Gain | AKAP13 LOC642423 LOC101929479 GOLGA6L3 MIR7706 |
nsv570298 | Chr.15:85272357 - 85510128 on Build GRCh38 | Gain | AKAP13 MIR7706 |
nsv428311 | Chr.15:85107707 - 85486274 on Build GRCh38 | Gain | AKAP13 LOC642423 LOC101929479 GOLGA6L3 MIR7706 PDE8A |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |