Assay Details
Target Gene Details
Entrez Gene ID: | 10519 |
Gene Name: | calcium and integrin binding 1 |
Gene Aliases: |
CIB, CIBP, KIP1, PRKDCIP, SIP2-28 |
Location: |
Chr.15:90230245-90265759 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CIB1 | NR_102428.1 | |||
BG759961.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833087 | Chr.15:90208675 - 90425725 on Build GRCh38 | Loss | IQGAP1 GDPGP1 NGRN SEMA4B GABARAPL3 CIB1 LOC400446 ZNF774 TTLL13P |
nsv1052652 | Chr.15:90101186 - 90298961 on Build GRCh38 | Gain | GDPGP1 IDH2 NGRN SEMA4B CIB1 LOC400446 TTLL13P |
dgv112e55 | Chr.15:90079272 - 90249274 on Build GRCh38 | Gain | GDPGP1 IDH2 ZNF710 SEMA4B CIB1 |
esv2760054 | Chr.15:90022724 - 90266401 on Build GRCh38 | Gain | GDPGP1 IDH2 ZNF710 NGRN SEMA4B CIB1 TTLL13P |
esv3637216 | Chr.15:90242762 - 90247368 on Build GRCh38 | Gain | CIB1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |