Assay Details
Target Gene Details
Entrez Gene ID: | 1543 |
Gene Name: | cytochrome P450 family 1 subfamily A member 1 |
Gene Aliases: |
AHH, AHRR, CP11, CYP1, CYPIA1, P1-450, P450-C, P450DX |
Location: |
Chr.15:74719542-74725610 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CYP1A1 | NM_000499.4 | NP_000490.1 | ||
NM_001319216.1 | NP_001306145.1 | |||
NM_001319217.1 | NP_001306146.1 | |||
XM_017021953.1 | XP_016877442.1 | |||
AK223108.1 | BAD96828.1 | |||
AK223113.1 | BAD96833.1 | |||
AK310810.1 | ||||
AK313880.1 | ||||
AM233517.1 | ||||
AM233518.1 | ||||
AM233519.1 | ||||
AM233520.1 | ||||
AM236046.1 | ||||
AM236047.1 | ||||
AY871801.1 | ||||
BC023019.1 | AAH23019.1 | |||
DA693879.1 | ||||
K03191.1 | AAA52139.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833055 | Chr.15:74696003 - 74873947 on Build GRCh38 | Loss | CPLX3 MIR4513 SCAMP2 ULK3 MIR6882 EDC3 CYP1A1 CSK CYP1A2 LMAN1L |
esv3636907 | Chr.15:74721538 - 74745196 on Build GRCh38 | Loss | CYP1A1 |
nsv817698 | Chr.15:74596822 - 74727108 on Build GRCh38 | Gain | ARID3B EDC3 CYP1A1 LOC102723750 CLK3 |
esv2763106 | Chr.15:74722123 - 74741845 on Build GRCh38 | Loss | CYP1A1 |
nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion | LOC105376731 MPI CYP11A1 NPTN SCAMP5 LOC283731 COX5A CD276 LMAN1L UBL7-AS1 RPP25 CPLX3 ADPGK FAM219B PML SEMA7A LOC101929333 NPTN-IT1 LOXL1 CYP1A2 MIR4513 C15orf59 ARID3B BBS4 CCDC33 PPCDC C15orf39 HIGD2B ADPGK-AS1 MIR6882 EDC3 GOLGA6A CYP1A1 LOC102723750 C15orf59-AS1 REC114 LOXL1-AS1 NEO1 ULK3 UBL7 ISLR2 HCN4 STRA6 GOLGA6C LOC729739 SCAMP2 STOML1 LOC107984728 LOC732265 MIR6881 CSK TBC1D21 ISLR CLK3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |