Assay Details
Target Gene Details
Entrez Gene ID: | 644192 |
Gene Name: | NR2F2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.15:96127360-96327361 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NR2F2-AS1 | NR_125738.1 | |||
DA941342.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1141674 | Chr.15:95302056 - 96435607 on Build GRCh38 | Deletion | NR2F2-AS1 NR2F2 LINC00924 LINC01197 MIR1469 |