Assay Details
Target Gene Details
Entrez Gene ID: | 5151 |
Gene Name: | phosphodiesterase 8A |
Gene Aliases: |
HsT19550 |
Location: |
Chr.15:84980513-85139145 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PDE8A | NM_001243137.1 | NP_001230066.1 | ||
NM_002605.2 | NP_002596.1 | |||
NM_173454.1 | NP_775656.1 | |||
XM_017022308.1 | XP_016877797.1 | |||
XM_017022309.1 | XP_016877798.1 | |||
XM_017022310.1 | XP_016877799.1 | |||
XM_017022311.1 | XP_016877800.1 | |||
AF332653.1 | AAK57641.1 | |||
AF388183.1 | AAL18610.1 | |||
AF388184.1 | AAL18611.1 | |||
AF388185.1 | AAL18612.1 | |||
AF388186.1 | AAL18613.1 | |||
AF388187.1 | AAL18614.1 | |||
AK074280.1 | ||||
BC060762.1 | AAH60762.1 | |||
BC075822.1 | AAH75822.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1040994 | Chr.15:84992252 - 85041926 on Build GRCh38 | Gain | PDE8A |
esv2672879 | Chr.15:84183834 - 85188520 on Build GRCh38 | Deletion | LOC105376722 ZNF592 UBE2Q2L UBE2Q2P1 LOC102724135 GOLGA2P7 GOLGA6L4 LOC440300 ALPK3 SEC11A CSPG4P5 ZSCAN2 WDR73 PDE8A LINC00933 NMB LOC103171574 SLC28A1 GOLGA6L5P SCAND2P |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |