Assay Details
Target Gene Details
Entrez Gene ID: | 90381 |
Gene Name: | TOPBP1 interacting checkpoint and replication regulator |
Gene Aliases: |
C15orf42, SLD3, Treslin |
Location: |
Chr.15:89575587-89628023 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TICRR | NM_001308025.1 | NP_001294954.1 | ||
NM_152259.3 | NP_689472.3 | |||
DR731357.1 | ||||
GQ227787.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3892721 | Chr.15:89530925 - 89673865 on Build GRCh38 | Gain | PLIN1 TICRR KIF7 |
nsv1038140 | Chr.15:89141274 - 89620108 on Build GRCh38 | Gain | POLG RLBP1 MIR6766 TICRR MIR9-3HG FANCI RHCG LOC105371031 LOC105370962 MIR9-3 ABHD2 LINC00928 |
esv34025 | Chr.15:89412131 - 89630626 on Build GRCh38 | Loss | TICRR RHCG LOC105371031 KIF7 LINC00928 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |