Assay Details
Target Gene Details
Entrez Gene ID: | 9154 |
Gene Name: | solute carrier family 28 member 1 |
Gene Aliases: |
CNT1, HCNT1 |
Location: |
Chr.15:84884351-84975649 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv524050 | Chr.15:84864333 - 84935498 on Build GRCh38 | Loss | SLC28A1 ALPK3 |
nsv471257 | Chr.15:84869224 - 84919085 on Build GRCh38 | Gain | SLC28A1 ALPK3 |
esv2672879 | Chr.15:84183834 - 85188520 on Build GRCh38 | Deletion | SEC11A GOLGA6L4 LINC00933 GOLGA2P7 ZSCAN2 LOC102724135 SCAND2P UBE2Q2L SLC28A1 NMB LOC103171574 LOC440300 CSPG4P5 WDR73 UBE2Q2P1 ZNF592 GOLGA6L5P ALPK3 PDE8A LOC105376722 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |