Assay Details
Target Gene Details
Entrez Gene ID: | 4016 |
Gene Name: | lysyl oxidase like 1 |
Gene Aliases: |
LOL, LOXL |
Location: |
Chr.15:73926451-73952141 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOXL1 | NM_005576.3 | NP_005567.2 | ||
XM_011521555.1 | XP_011519857.1 | |||
AK303982.1 | ||||
AK314222.1 | ||||
BC015090.2 | AAH15090.1 | |||
BC068542.1 | AAH68542.1 | |||
L21186.1 | AAA50162.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1603 | Chr.15:73898881 - 73943392 on Build GRCh38 | Deletion | LOXL1-AS1 TBC1D21 LOXL1 |
esv2760040 | Chr.15:73922773 - 74172575 on Build GRCh38 | Gain+Loss | ISLR2 GOLGA6A PML LOXL1-AS1 LOXL1 LOC732265 LOC283731 STOML1 |
nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion | C15orf39 LOC729739 CPLX3 SCAMP5 BBS4 EDC3 ADPGK-AS1 NEO1 NPTN-IT1 PML SCAMP2 LOC101929333 CD276 NPTN CSK GOLGA6A TBC1D21 ULK3 ARID3B LOC107984728 C15orf59 UBL7-AS1 LMAN1L REC114 MIR6881 CCDC33 ISLR LOXL1 LOC283731 STOML1 MPI UBL7 COX5A LOXL1-AS1 ADPGK SEMA7A MIR6882 MIR4513 ISLR2 STRA6 HCN4 CYP1A1 RPP25 CLK3 FAM219B LOC105376731 LOC732265 GOLGA6C PPCDC CYP1A2 CYP11A1 LOC102723750 HIGD2B C15orf59-AS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |