Assay Details
Target Gene Details
Entrez Gene ID: | 57611 |
Gene Name: | immunoglobulin superfamily containing leucine rich repeat 2 |
Gene Aliases: |
LINX |
Location: |
Chr.15:74128203-74141826 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ISLR2 | NM_001130136.1 | 1 | 255 | NP_001123608.1 |
NM_001130137.1 | NP_001123609.1 | |||
XM_011521840.2 | XP_011520142.1 | |||
XM_011521841.1 | XP_011520143.1 | |||
AB040898.2 | 1 | 255 | ||
AK092302.1 | 1 | 271 | ||
BC152429.1 | 1 | 255 | ||
BI917487.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833053 | Chr.15:74067815 - 74262799 on Build GRCh38 | Loss | ISLR2 STRA6 CCDC33 LOC283731 ISLR GOLGA6A LOC732265 |
nsv1050884 | Chr.15:74102642 - 74197072 on Build GRCh38 | Gain | ISLR2 STRA6 LOC283731 ISLR |
nsv469619 | Chr.15:73999518 - 74173980 on Build GRCh38 | Loss | ISLR2 PML LOC283731 ISLR GOLGA6A LOC732265 |
nsv827385 | Chr.15:74127198 - 74136960 on Build GRCh38 | Gain | ISLR2 LOC283731 |
nsv951993 | Chr.15:74124460 - 74137359 on Build GRCh38 | Deletion | ISLR2 LOC283731 |
esv2760040 | Chr.15:73922773 - 74172575 on Build GRCh38 | Gain+Loss | STOML1 ISLR2 PML LOC283731 LOXL1 LOXL1-AS1 GOLGA6A LOC732265 |
nsv1047564 | Chr.15:74116495 - 74165840 on Build GRCh38 | Gain | ISLR2 LOC283731 |
nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion | SCAMP2 TBC1D21 HCN4 NEO1 FAM219B REC114 CSK STOML1 MIR6881 LOC101929333 CYP1A2 SEMA7A C15orf39 ADPGK-AS1 LOC729739 CCDC33 CPLX3 MIR4513 SCAMP5 HIGD2B COX5A PPCDC ISLR MIR6882 C15orf59 UBL7-AS1 NPTN LOXL1 LOXL1-AS1 NPTN-IT1 ARID3B STRA6 MPI GOLGA6A PML RPP25 CYP1A1 C15orf59-AS1 LOC105376731 GOLGA6C LOC732265 LMAN1L ISLR2 LOC102723750 EDC3 CD276 LOC283731 UBL7 ULK3 CYP11A1 LOC107984728 ADPGK CLK3 BBS4 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |