Assay Details
Target Gene Details
Entrez Gene ID: | 113146 |
Gene Name: | AHNAK nucleoprotein 2 |
Gene Aliases: |
C14orf78 |
Location: |
Chr.14:104937253-104978357 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AHNAK2 | NM_138420.2 | NP_612429.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1438 | Chr.14:104946391 - 104991244 on Build GRCh38 | Deletion | C14orf79 AHNAK2 |
nsv566141 | Chr.14:104938047 - 105089059 on Build GRCh38 | Gain | C14orf79 CDCA4 AHNAK2 GPR132 |
esv3635634 | Chr.14:104965985 - 105147574 on Build GRCh38 | Gain | LOC102723354 JAG2 C14orf79 CDCA4 AHNAK2 GPR132 |
nsv952276 | Chr.14:104922864 - 105021963 on Build GRCh38 | Deletion | C14orf79 CDCA4 PLD4 AHNAK2 |
nsv1049253 | Chr.14:104918622 - 105005518 on Build GRCh38 | Gain | C14orf79 PLD4 AHNAK2 |
nsv1054574 | Chr.14:103210111 - 105365114 on Build GRCh38 | Loss | LOC105378183 BTBD6 MIR203B INF2 C14orf180 JAG2 MIR6765 TRMT61A BAG5 LINC00638 XRCC3 C14orf79 TMEM179 RD3L LINC00637 GPR132 CKB APOPT1 NUDT14 SIVA1 KLC1 ASPG LOC388022 MARK3 MIR203A ZFYVE21 LOC105370708 TDRD9 BRF1 PACS2 MIR4710 AHNAK2 CEP170B EIF5 PPP1R13B LOC102723354 SNORA28 LOC107984648 ADSSL1 KIF26A ZBTB42 AKT1 C14orf2 LOC105370687 LOC101929634 CDCA4 LOC105370691 PLD4 |
nsv1052190 | Chr.14:104957424 - 105256316 on Build GRCh38 | Gain | BTBD6 LOC102723354 JAG2 MIR6765 C14orf79 BRF1 CDCA4 AHNAK2 GPR132 NUDT14 |
nsv566150 | Chr.14:104969312 - 105223755 on Build GRCh38 | Loss | LOC102723354 JAG2 MIR6765 C14orf79 BRF1 CDCA4 AHNAK2 GPR132 NUDT14 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |